Skip to Content
Merck
All Photos(4)

Key Documents

D2521

Sigma-Aldrich

(+)-cis-Diltiazem hydrochloride

≥99% (HPLC)

Synonym(s):

(2S,3S)-(+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one hydrochloride, CRD-401

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O4S · HCl
CAS Number:
Molecular Weight:
450.98
Beilstein:
4228706
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

powder

originator

Valeant

storage temp.

2-8°C

SMILES string

Cl.COc1ccc(cc1)[C@@H]2Sc3ccccc3N(CCN(C)C)C(=O)[C@@H]2OC(C)=O

InChI

1S/C22H26N2O4S.ClH/c1-15(25)28-20-21(16-9-11-17(27-4)12-10-16)29-19-8-6-5-7-18(19)24(22(20)26)14-13-23(2)3;/h5-12,20-21H,13-14H2,1-4H3;1H/t20-,21+;/m1./s1

InChI key

HDRXZJPWHTXQRI-BHDTVMLSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diltiazem hydrochloride is used to treat cardiovascular diseases, such as angina and hypertension. It acts as an inhibitor to cytochrome P450 3A (CYP3A). Diltiazem might be associated with maculopapular rashes, urticaria and pruritus.

Application

(+)-cis-Diltiazem hydrochloride has been used:
  • to study its effects on diffusion and release in monolithic dispersion matrix(26)
  • to study its neuroprotective effects in retinitis pigmentosa (RP) models(27)
  • as an antagonist against the Cav1.2 L-type calcium channels (LTCCs)(28)

Biochem/physiol Actions

Blocks slow, or L-type Ca2+ channels; regulates Ca2+ release from intracellular stores in neutrophils; stimulates 1,4-dihydropyridine binding to Ca2+ channels; coronary vasodilator.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Calcium Channels and Cyclic Nucleotide-Gated (CNG) and Hyperpolarization Activated Cyclic Nucleotide-Gated (HCN) Channels pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Valeant. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diltiazem induces severe photodistributed hyperpigmentation: case series, histoimmunopathology, management, and review of the literature
Saladi RN, et al.
Archives of Dermatology, 142(2), 206-210 (2006)
D Dobrev et al.
British journal of pharmacology, 127(2), 576-582 (1999-06-29)
1. The putative inhibitory effects of verapamil and diltiazem on neuronal non-L-type Ca2+ channels were studied by investigating their effects on either K+- or veratridine-evoked [3H]-dopamine ([3H]-DA) release in rat striatal slices. Involvement of N-, P- and Q-type channels was
Andrés Pizzorno et al.
Cell reports. Medicine, 1(4), 100059-100059 (2020-08-25)
In the current COVID-19 pandemic context, proposing and validating effective treatments represents a major challenge. However, the scarcity of biologically relevant pre-clinical models of SARS-CoV-2 infection imposes a significant barrier for scientific and medical progress, including the rapid transition of
Effect of diltiazem on midazolam and alfentanil disposition in patients undergoing coronary artery bypass grafting
Ahonen J, et al.
Anesthesiology, 85(6), 1246-1252 (1996)
Determination of plasma protein binding of diltiazem in horses by ultrafiltration.
C C Schwarzwald et al.
Journal of veterinary pharmacology and therapeutics, 29(6), 579-580 (2006-11-07)

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service