
4-Nitrophenyl β-D-mannopyranoside

Synonym(s):
p-Nitrophenyl β-D-mannopyranoside
Empirical Formula (Hill Notation):
C12H15NO8
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing