Skip to Content
Merck
All Photos(1)

Key Documents

N1016

Sigma-Aldrich

Nimesulide

Synonym(s):

N-(4-Nitro-2-phenoxyphenyl)methanesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N2O5S
CAS Number:
Molecular Weight:
308.31
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

ethanol: 50 mg/mL

storage temp.

2-8°C

SMILES string

CS(NC1=C(OC2=CC=CC=C2)C=C([N+]([O-])=O)C=C1)(=O)=O

InChI

1S/C13H12N2O5S/c1-21(18,19)14-12-8-7-10(15(16)17)9-13(12)20-11-5-3-2-4-6-11/h2-9,14H,1H3

InChI key

HYWYRSMBCFDLJT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Nimesulide, a nonsteroidal anti-inflammatory drug (NSAID) belongs to the sulfonanilide class.[1]

Application

Nimesulide has been used as a component in the reaction mixture to determine the activity of cyclooxygenase (COX) in spleen of chickens.[2]

Biochem/physiol Actions

Highly selective cyclooxygenase-2 inhibitor.
Nimesulide can be used as a substitute to other nonsteroidal anti-inflammatory drugs (NSAIDs) to treat pain and inflammation of osteoarthritis for a short period. It can perform anti-inflammatory, analgesic and antipyretic activities.[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nimesulide
Davis R and Brogden R N
Drugs, 48(3), 431-454 (1994)
Effects of dietary fish oil on NF
Yang X, et al.
The Journal of animal ecology, 20(3), 206-223 (2011)
Renata P Raffin et al.
Journal of biomedical nanotechnology, 8(2), 309-315 (2012-04-21)
Lipid nanoparticles are drug delivery systems able to increase bioavailability of poorly soluble drugs. They can be prepared with different lipid materials, especially natural lipids. Shea butter is a natural lipid obtained from the Butyrospermum parkii seed and rich in
Brijesh Kumar Singh et al.
PloS one, 7(4), e34200-e34200 (2012-04-18)
Nimesulide, an anti-inflammatory and analgesic drug, is reported to cause severe hepatotoxicity. In this study, molecular mechanisms involved in deranged oxidant-antioxidant homeostasis and mitochondrial dysfunction during nimesulide-induced hepatotoxicity and its attenuation by plant derived terpenes, camphene and geraniol has been
Yuan Yuan et al.
International journal of pharmaceutics, 430(1-2), 114-119 (2012-04-17)
Poloxamer 407 has excellent thermo-sensitive gelling properties. Nevertheless, these gels possess inadequate poor bioadhesiveness and high permeability to water, which limited its' application as a thermoresponsive matrix. The main aim of the present investigation was to develop thermosensitive and mucoadhesive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service