Skip to Content
Merck
All Photos(1)

Key Documents

N0845000

Nimesulide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-(4-Nitro-2-phenoxyphenyl)methanesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N2O5S
CAS Number:
Molecular Weight:
308.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nimesulide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CS(NC1=C(OC2=CC=CC=C2)C=C([N+]([O-])=O)C=C1)(=O)=O

InChI

1S/C13H12N2O5S/c1-21(18,19)14-12-8-7-10(15(16)17)9-13(12)20-11-5-3-2-4-6-11/h2-9,14H,1H3

InChI key

HYWYRSMBCFDLJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nimesulide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Highly selective cyclooxygenase-2 inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shannon L Dean et al.
Endocrinology, 153(11), 5415-5427 (2012-10-12)
Prostaglandins (PGs) are ubiquitous membrane-derived, lipid-signaling molecules with wide ranging effects throughout the body. In the brain, PGE(2) is the key regulator of fever after inflammation but is also implicated in neural development and synaptic plasticity. The steroid hormone estradiol
Yuan Yuan et al.
International journal of pharmaceutics, 430(1-2), 114-119 (2012-04-17)
Poloxamer 407 has excellent thermo-sensitive gelling properties. Nevertheless, these gels possess inadequate poor bioadhesiveness and high permeability to water, which limited its' application as a thermoresponsive matrix. The main aim of the present investigation was to develop thermosensitive and mucoadhesive
Brijesh Kumar Singh et al.
PloS one, 7(4), e34200-e34200 (2012-04-18)
Nimesulide, an anti-inflammatory and analgesic drug, is reported to cause severe hepatotoxicity. In this study, molecular mechanisms involved in deranged oxidant-antioxidant homeostasis and mitochondrial dysfunction during nimesulide-induced hepatotoxicity and its attenuation by plant derived terpenes, camphene and geraniol has been
Renata P Raffin et al.
Journal of biomedical nanotechnology, 8(2), 309-315 (2012-04-21)
Lipid nanoparticles are drug delivery systems able to increase bioavailability of poorly soluble drugs. They can be prepared with different lipid materials, especially natural lipids. Shea butter is a natural lipid obtained from the Butyrospermum parkii seed and rich in
Y Chai et al.
British journal of cancer, 108(5), 1106-1112 (2013-02-16)
Previous studies from our group and others have shown that cyclooxygenase-2 (COX-2) has an essential role in radiation-induced non-targeted responses and genomic instability in vivo. However, the signalling pathways involved in such effects remain unclear. A 1 cm(2) area (1 cm ×

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service