Skip to Content
Merck
All Photos(1)

Key Documents

Q1125

Sigma-Aldrich

Quinine hydrochloride dihydrate

99-101% (titration), potassium channel blocker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2 · HCl · 2H2O
CAS Number:
Molecular Weight:
396.91
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.77

product name

Quinine hydrochloride dihydrate,

Quality Level

solubility

H2O: 0.25/5 g/mL, clear, colorless

antibiotic activity spectrum

parasites

Mode of action

enzyme | inhibits

SMILES string

O.Cl.[H][C@]1(C[C@@H]2CC[N@H]1C[C@@H]2C=C)[C@H](O)c3ccnc4ccc(OC)cc34

InChI

1S/C20H24N2O2.ClH.H2O/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;;/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;1H;1H2/t13-,14-,19-,20+;;/m0../s1

InChI key

SGVZDMWHXVXUBY-HZQSTTLBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quinine is derived from the stem of cinchona (quina-quina) tree. It is an alkaloid having various applications in the field of medicine. Quinine is a basic compound with aryl amino alcohol group.

Application

Quinine hydrochloride dihydrate has been used in Drosophila larval behavioral assay. It has also been used in taste preference analysis among rats.

Biochem/physiol Actions

Quinine is widely used in the treatment of malaria. It possesses erythrocytic and gametocytocidal action against Plasmodium sp. It also serves as an analgesic drug. Upon administration, quinine is present mostly bound to α-1 acid glycoprotein in the body fluids. Quinine has the ability to cross the placenta and the blood brain barrier. It has a half-life of 11-18 hours. Tinnitus, hearing defects, nausea and headache are some of its side effects.
Potassium channel blocker

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-NaCl perception in Drosophila melanogaster
Alves G, et al.
The Journal of Neuroscience, 34(33), 10884-10891 (2014)
The first stereoselective total synthesis of quinine
Stork G, et al.
Journal of the American Chemical Society, 123(14), 3239-3242 (2001)
Preferences of 14 rat strains for 17 taste compounds
Tordoff MG, et al.
Physiology & Behavior, 95(3), 308-332 (2008)
Eleanor K Lutz et al.
The Journal of experimental biology, 223(Pt 7) (2020-03-05)
Mosquitoes spread deadly diseases that impact millions of people every year. Understanding mosquito physiology and behavior is vital for public health and disease prevention. However, many important questions remain unanswered in the field of mosquito neuroethology, particularly in our understanding
Hu Chen et al.
Addiction biology, 25(6), e12821-e12821 (2019-08-23)
One of the most pernicious characteristics of alcohol use disorder is the compulsion to drink despite negative consequences. The insular cortex controls decision making under conditions of risk or conflict. Cortical activity is tightly controlled by inhibitory interneurons that are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service