Skip to Content
Merck
All Photos(2)

Key Documents

M5883

Sigma-Aldrich

7-Methylguanosine 5′-diphosphate sodium salt

≥92.5%

Synonym(s):

7-Methyl-gdp, m7GDP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17N5O11P2 · xNa+
CAS Number:
Molecular Weight:
457.23 (free acid basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥92.5%

form

powder

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

[Na+].[Na+].C[n+]1cn([C@@H]2O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]2O)c3nc(N)nc([O-])c13

InChI

1S/C11H17N5O11P2.2Na/c1-15-3-16(8-5(15)9(19)14-11(12)13-8)10-7(18)6(17)4(26-10)2-25-29(23,24)27-28(20,21)22;;/h3-4,6-7,10,17-18H,2H2,1H3,(H5-,12,13,14,19,20,21,22,23,24);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1

InChI key

VSMXIKIFPKDZJA-IDIVVRGQSA-L

General description

7-Methylguanosine 5′-diphosphate (m7GDP) is derived when the human decapping Dcp2 enzyme cleaves the cap of the deadenylated mRNAs during the 5′–3′ mRNA decay pathway.

Application

7-Methylguanosine 5′-diphosphate sodium salt has been used as a component of the elution buffer for the purification of 5’capped RNA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Długosz et al.
Journal of biochemical and biophysical methods, 51(2), 179-193 (2002-06-14)
A method for extracting kinetic and optical parameters from progress curves for protein-ligand association, obtained by stopped-flow experiments, is described. The method is limited to one-step and two-step association kinetics, but it allows concentration of protein and offset of the
DcpS can act in the 5??3? mRNA decay pathway in addition to the 3??5? pathway
Erwin VD, et al.
Proceedings of the National Academy of Sciences, 100(21) (2003)
H Matsuo et al.
Nature structural biology, 4(9), 717-724 (1997-09-26)
eIF4E, the mRNA cap binding protein, is a master switch that controls eukaryotic translation. To be active, it must bind eIF4G and form the eIF4F complex, which also contains eIF4A. Translation is downregulated by association of eIF4E with 4E-BP, which
A B Sachs et al.
Nature structural biology, 7(5), 356-361 (2000-05-10)
The eukaryotic cap and poly(A) tail binding proteins, eIF4E and Pab1p, play important roles in the initiation of protein synthesis. The recent structures of the complex of eIF4E bound to the methylated guanosine (cap) found at the 5'end of messenger
7-methylguanosine diphosphate (m7GDP) is not hydrolyzed but strongly bound by Decapping Scavenger (DcpS) enzymes and potently inhibits their activity
Wypijewska A, et al.
Biochemistry, 51(40) (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service