Skip to Content
Merck
All Photos(7)

Key Documents

I202

Sigma-Aldrich

Imidazole

ReagentPlus®, 99%

Synonym(s):

1,3-Diaza-2,4-cyclopentadiene, Glyoxaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Beilstein:
103853
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
39161001
PubChem Substance ID:
NACRES:
NA.25

vapor pressure

<1 mmHg ( 20 °C)

Quality Level

product line

ReagentPlus®

Assay

99%

form

crystalline

pH

9-11 at 100 g/L (23 °C)

pKa (25 °C)

6.95

bp

256 °C (lit.)

mp

88-91 °C (lit.)

SMILES string

c1c[nH]cn1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Excellent for buffers in the range of pH 6.2-7.8
Imidazole is a biocompatible molecule that is used as a scaffold for biomimetic applications. It is used as a bioreagent with an ability to hydrogen bond with drugs and proteins. It can be also be used as an additive for the formation of an electrolyte for fuel cell applications.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imidazole and 1-methyl imidazole in phosphoric acid doped polybenzimidazole, electrolyte for fuel cells
Schechter A and Savinell RF
Solid State Ionics, 147(1-2), 181-187 (2002)
Three-Dimensional Covalent Co-Assembly between Inorganic Supertetrahedral Clusters and Imidazolates.
Wu T, et al.
Angewandte Chemie (International Edition in English), 50(11), 2536-2539 (2011)
Imidazole-and imidazolium-containing polymers for biology and material science applications
Anderson EB and Long TE
Polymer, 51(12), 2447-2454 (2010)
Cobalt imidazolate framework as precursor for oxygen reduction reaction electrocatalysts.
Ma S, et al.
Chemistry?A European Journal , 17(7), 2063-2067 (2011)
Ali Tebbi et al.
The Journal of biological chemistry, 290(22), 14077-14090 (2015-04-17)
Ribonucleotide reductase (RnR) is a key enzyme synthesizing deoxyribonucleotides for DNA replication and repair. In mammals, the R1 catalytic subunit forms an active complex with either one of the two small subunits R2 and p53R2. Expression of R2 is S

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Related Content

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service