All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H15N5O5
CAS Number:
Molecular Weight:
297.27
Beilstein:
3713683
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Recommended Products
biological source
synthetic
Quality Level
Assay
≥90%
form
powder
solubility
water: 50 mg/mL, clear to hazy, colorless to faintly yellow
storage temp.
2-8°C
SMILES string
C[n+]1cn([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)c3nc(N)nc([O-])c13
InChI
1S/C11H15N5O5/c1-15-3-16(8-5(15)9(20)14-11(12)13-8)10-7(19)6(18)4(2-17)21-10/h3-4,6-7,10,17-19H,2H2,1H3,(H2-,12,13,14,20)/t4-,6-,7-,10-/m1/s1
InChI key
OGHAROSJZRTIOK-KQYNXXCUSA-N
Looking for similar products? Visit Product Comparison Guide
General description
7-Methylguanosine structure is also referred to as a cap. It is located on the 5′ end of the cytoplasmic mRNA.
Application
7-Methylguanosine has been used in the removal of residual phosphates from pre rigor solution, phosphate mop system and phosphorous standard solution.
Biochem/physiol Actions
7-Methylguanosine is involved in the processing of a cap-dependent translational process of mRNA. It is also involved in protecting the mRNA from degradation due to ribonucleases and mediate nuclear export. 7-Methylguanosine also permits the recruitment of ribosome.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Vishnu Modur et al.
Nature communications, 9(1), 4410-4410 (2018-10-26)
The nature and role of global transcriptional deregulations in cancers are not fully understood. We report that a large proportion of cancers have widespread defects in mRNA transcription elongation (TE). Cancers with TE defects (TEdeff) display spurious transcription and defective
Nicolas Leulliot et al.
Structure (London, England : 1993), 16(1), 52-61 (2008-01-11)
Loss of N7-methylguanosine (m7G) modification is involved in the recently discovered rapid tRNA degradation pathway. In yeast, this modification is catalyzed by the heterodimeric complex composed of a catalytic subunit Trm8 and a noncatalytic subunit Trm82. We have solved the
Akiko Ogawa et al.
STAR protocols, 2(4), 100848-100848 (2021-10-09)
About 150 modifications have been identified in RNA species. Besides their regulatory roles in the intracellular gene expression, abundant modified RNA nucleosides are catabolized from RNA and released into extracellular fluids, which can impact extracellular signaling as ligands for receptors.
Jinpeng Gao et al.
Plant physiology, 174(3), 1713-1727 (2017-05-13)
Chilling stress is a major factor limiting plant development and crop productivity. Because the plant response to chilling is so complex, we are far from understanding the genes important in the response to chilling. To identify new genes important in
G Hu et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(13), 7149-7154 (1999-06-23)
We have determined, by high resolution x-ray analysis, 10 structures comprising the mRNA cap-specific methyltransferase VP39 or specific mutants thereof in the presence of methylated nucleobase analogs (N1-methyladenine, N3-methyladenine, N1-methylcytosine, N3-methylcytosine) and their unmethylated counterparts, or nucleoside N7-methylguanosine. Together with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service