Skip to Content
Merck
All Photos(4)

Key Documents

N2415

Sigma-Aldrich

Nucleoside Phosphorylase bacterial

recombinant, expressed in E. coli, ≥10 units/mg protein

Synonym(s):

PNP, Purine nucleoside phosphorylase, Purine nucleoside:orthophosphate ribosyltransferase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

Assay

≥80% protein basis (biuret)

form

lyophilized powder

specific activity

≥10 units/mg protein

sequence note

MATPHINAEMGDFADVVLMPGDPLRAKYIAETFLEDAREVNNVRGMLGFTGTYKGRKISVMGHGMGIPSCSIYTKELITDFGVKKIIRVGSCGAVLPHVKLRDVVIGMGACTDSKVNRIRFKDHDFAAIADFDMVRNAVDAAKALGIDARVGNLFSADLFYSPDGEMFDVMEKYGILGVEMEAAGIYGVAAEFGAKALTICTVSDHIRTHEQTTAAERQTTFNDMIKIALESVLLGDKE

UniProt accession no.

storage temp.

−20°C

SMILES string

[n]2(c3ncncc3nc2)[C@@H]1O[C@@H]([C@H]([C@H]1O)O)CO

InChI

1S/C10H12N4O4/c15-2-6-7(16)8(17)10(18-6)14-4-13-5-1-11-3-12-9(5)14/h1,3-4,6-8,10,15-17H,2H2/t6-,7-,8-,10-/m1/s1

InChI key

MRWXACSTFXYYMV-FDDDBJFASA-N

Gene Information

Escherichia coli ... deoD(945654)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Bacterial nucleoside phosphorylase has been used in a study that identified and characterized two adenosine phosphorylase activities in Mycobacterium smegmatis. Bacterial nucleoside phosphorylase has also been used in a study to investigate the inhibitionof pyrimidine and purine nucleoside phosphorylases by a 3,5-dichlorobenzoyl-substituted 2-deoxy-D-ribose-1-phosphate derivative.
Nucleoside Phosphorylase bacterial has been used in reaction buffer and buffer-exchanged to remove residual phosphate.
Nucleoside phosphorylase is used in coupled enzyme systems to measure protein dephosphorylation.

Biochem/physiol Actions

Catalyzes the following reaction:purine nucleoside + phosphate = purine + alpha-D-ribose 1-phosphate
Nucleoside Phosphorylase/purine nucleoside phosphorylase (PNP), an enzyme participates in the purine salvage pathway. It also helps in the reversible cleavage of inosine to hypoxanthine and guanosine to guanine. PNP deficiency results in neurologic manifestations.

Unit Definition

One unit will cause the phosphorolysis of 1.0 micromole of inosine to hypoxanthine andribose-1-phosphate per min at pH 7.4 at 25 deg C.

Preparation Note

This is a salt-free and animal component-free enzyme preparation. This enzyme should be solubilized in a 10mM KPO4, pH 7.4 solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Primary T-cell immunodeficiencies
Clinical Immunology (Orlando, Fla.), 54(7), 489-508 (2019)
Disorders of purines and pyrimidines
Handbook of Clinical Neurology, 827-838 (2014)
Conformational stability and catalytic activity of PTEN variants linked to cancers and autism spectrum disorders
Johnston S B and Raines R T
Biochemistry, 54(7), 1576-1582 (2015)
Patricia Calero et al.
Nature communications, 11(1), 5045-5045 (2020-10-09)
Fluorine is a key element in the synthesis of molecules broadly used in medicine, agriculture and materials. Addition of fluorine to organic structures represents a unique strategy for tuning molecular properties, yet this atom is rarely found in Nature and
Irina V Varizhuk et al.
Current protocols, 2(1), e347-e347 (2022-01-21)
A simple and efficient method for the preparation of α-D-ribose 1-phosphate and 2-deoxy-α-D-ribose 1-phosphate, key intermediates in nucleoside metabolism and important starting compounds for the enzymatic synthesis of various modified nucleosides, has been proposed. It consists in near-irreversible enzymatic phosphorolysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service