Skip to Content
Merck
All Photos(1)

Key Documents

L4395

Sigma-Aldrich

Liposome Kit: Lipid mixtures for the preparation of liposomes

lyophilized powder

Synonym(s):

cholesterol, L-α-phosphatidylcholine and stearylamine kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

egg yolk

Quality Level

form

lyophilized powder

composition

Cholesterol, 9 μmol/package
L-α-Phosphatidylcholine (egg yolk), 63 μmol/package
Stearylamine, 18 μmol/package

functional group

carboxylic acid

shipped in

ambient

storage temp.

−20°C

Related Categories

Application

Liposome Kit has been used for the preparation of liposomes.

Biochem/physiol Actions

Liposome Kit: Lipid mixtures for the preparation of liposomes contains cholesterol; L-α-phosphatidylcholine and stearylamine. Liposome kit may be used to prepare cationic (positively charge) liposomes for incorporation of materials into cells.
Liposomes can be used for different applications such as drug delivery, diagnostics, membrane models, adjuvants for vaccination and others. This lipid mixture can be used to encapsulate a broad spectrum of hydrophilic and amphipathic molecules of low, medium, and high molecular weight (including peptides, proteins, and oligo- and polynucleotides).

related product

Product No.
Description
Pricing

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santosh Kumar et al.
JCI insight, 6(4) (2021-01-15)
Prime-boost immunization strategies are required to control the global tuberculosis (TB) pandemic, which claims approximately 3 lives every minute. Here, we have generated an immunogenic complex against Mycobacterium tuberculosis (M.tb), consisting of promiscuous T cell epitopes (M.tb peptides) and TLR
Xuan T A Nguyen et al.
Molecular neurobiology, 56(9), 6121-6133 (2019-02-08)
The cellular prion protein (PrPC), mainly known for its role in neurodegenerative diseases, is involved in several physiological processes including neuritogenesis. In addition, its ability to bind copper or zinc has been suggested for its role in metal homeostasis. Although
Inhibition of cationic liposomes of [3H]thymidine incorporation into DNA of L1210 cells.
Laurent G, Laduron C, Ruysschaert JM, et al.
Research Communications in Molecular Pathology and Pharmacology, 31, 515-527 (1981)
Abd Kakhar Umar et al.
Molecules (Basel, Switzerland), 26(17) (2021-09-11)
Human epidermal growth factor (hEGF) has been known to have excellent wound-healing activity. However, direct application to the wound area can lead to low hEGF bioavailability due to protease enzymes or endocytosis. The use of liposomes as coatings and carriers
Sub-Micrometric Liposomes as Drug Delivery Systems in the Treatment and Periodontitis
Di Turi G, et al.
International Journal of Immunopathology and Pharmacology, 25(3), 657-670 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service