Skip to Content
Merck
All Photos(1)

Key Documents

L5146

Sigma-Aldrich

Lipid Mixture (1000×)

liquid, sterile-filtered, BioReagent, suitable for insect cell culture

Synonym(s):

Lipid Mixture for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

product line

BioReagent

form

liquid

composition

cholesterol, 4.5 g/L
cod liver oil fatty acids (methyl esters), 10 g/L
polyoxyethylenesorbitan monooleate, 25 g/L
D-α-tocopherol acetate, 2.0 g/L

technique(s)

cell culture | insect: suitable

storage temp.

2-8°C

Application

Lipid Mixture (1000×) is used as a component in culture medium (of Perkinsus marinus strain). It is also used in Perkinsus marinus metabolite labeling and analysis.

Biochem/physiol Actions

The density of a lipid mixture is composed of nearly 85% wax esters. The wax ester mixture melts as a function of temperature at atmospheric pressure. No significant indications of non-random long-range lateral lipid distribution are seen in the all-fluid and all-gel state of the lipid mixture at ambient pressure. The morphology of the concentration fluctuations that is observed within the two-phase coexistence region of the lipid mixture can be characterized as a complex heterogeneous network of coexisting clusters with fractal-like properties.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isao Masuda et al.
Nucleic acids research, 38(18), 6186-6194 (2010-05-29)
Diverse mitochondrial (mt) genetic systems have evolved independently of the more uniform nuclear system and often employ modified genetic codes. The organization and genetic system of dinoflagellate mt genomes are particularly unusual and remain an evolutionary enigma. We determined the
A Pestronk et al.
Neurology, 48(4), 1104-1106 (1997-04-01)
IgM anti-GM1 antibodies are associated with motor neuropathy syndromes, including multifocal motor neuropathy (MMN). We compared the ability of serum IgM from patients with multifocal motor neuropathy to bind to GM1 ganglioside alone and to GM1 as a component of
Kristin Noell et al.
Omics : a journal of integrative biology, 25(2), 93-101 (2021-02-12)
Perkinsus marinus, a protozoan and the causative agent of perkinsosis (dermo disease) is a prevalent parasite found within the eastern oyster (Crassostrea virginica). In this study, we explore metabolic processes of P. marinus cells under lipid-depleted medium conditions to elucidate
Jillian C Danne et al.
Molecular biology and evolution, 30(1), 123-139 (2012-08-28)
Mitochondrial metabolism is central to the supply of ATP and numerous essential metabolites in most eukaryotic cells. Across eukaryotic diversity, however, there is evidence of much adaptation of the function of this organelle according to specific metabolic requirements and/or demands
Motomichi Matsuzaki et al.
Molecular biology and evolution, 25(6), 1167-1179 (2008-03-25)
Plastids are widespread in plant and algal lineages. They are also exploited by some nonphotosynthetic protists, including malarial parasites, to support their diverse modes of life. However, cryptic plastids may exist in other nonphotosynthetic protists, which could be important in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service