Skip to Content
Merck
All Photos(1)

Key Documents

L3289

Sigma-Aldrich

Lysis solution for blood

sufficient for 100 reactions, for molecular biology

Synonym(s):

Blood direct PCR, Blood lysis buffer, Extract-N-Amp blood direct PCR lysis solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

grade

for molecular biology

Quality Level

form

liquid

usage

sufficient for 100 reactions

storage temp.

−20°C

General description

Lysis solution for blood in combination with neutralization solution for blood (N9784) aids in rapid extraction and neutralization of DNA from whole blood, whole blood dried on a blood card, and cultured mammalian cells.

This lysis solution is a component of the Extract-N-Amp and the REDExtract-N-Amp Blood PCR Kits

Application

Lysis solution for blood has been used for lysis of blood cells for DNA extraction prior to polymerase chain reaction (PCR) amplification. It has also been used for the isolation of genomic DNA for CRISPR-Cas9 gene editing.

Legal Information

Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification by random mutagenesis of functional domains in KREPB5 that differentially affect RNA editing between life cycle stages of Trypanosoma brucei
McDermott S M, et al.
Molecular and Cellular Biology, MCB-00790 (2015)
Identification by random mutagenesis of functional domains in KREPB5 that differentially affect RNA editing between life cycle stages of Trypanosoma brucei
McDermott S M, et al.
Molecular and cellular biology, MCB-00790 (2015)
Shuhan Chen et al.
Stem cell research, 45, 101804-101804 (2020-04-28)
Mutations in the Leucine rich repeat kinase 2 (LRRK2) gene are found in both familial and sporadic Parkinson's disease (PD), and are also associated with immune-related disorders including Crohn's disease (CD) and leprosy. We have generated two homozygous LRRK2 knockout
Leandro S Thiago et al.
Haematologica, 99(1), 155-162 (2013-07-23)
The B-cell compartment in which multiple myeloma stem cells reside remains unclear. We investigated the potential presence of mature, surface-membrane immunoglobulin-positive B lymphocytes clonally related to the tumor bone marrow plasma cells among different subsets of peripheral blood B cells
G Henderson et al.
Archives of disease in childhood. Fetal and neonatal edition, 94(2), F124-F128 (2007-09-05)
The inflammatory cytokine cascade is implicated in the pathogenesis of necrotising enterocolitis (NEC). Genetic association studies of cytokine polymorphisms may help to detect molecular mechanisms that are causally related to the disease process. To examine associations between the common genetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service