Skip to Content
Merck
All Photos(1)

Key Documents

N3910

Sigma-Aldrich

Neutralization Solution B

Synonym(s):

DNA neutralization solution, neutralization buffer DNA extraction

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161703
NACRES:
NA.25

grade

for molecular biology

Quality Level

form

liquid

technique(s)

DNA amplification: suitable
DNA extraction: suitable

storage temp.

−20°C

General description

This Neutralization Buffer B is intended for use with Sigma′s REDExtract-N-Amp Tissue PCR Kits. This buffer is used to neutralize the DNA extract and be combined with the REDExtract-N-Amp PCR Reaction Mix and PCR primers to amplify target DNA. The REDExtract-N-Amp Kits include a sufficient volume of Neutralization Buffer B for at least one neutralization per extraction. However, where additional amplifications are required, supplemental Neutralization buffer may be needed.

Application

Neutralization Solution B has been used in the modified Sigma Extract-N-Amp extraction procedure. It has also been used in genotyping of mice, for neutralizing the DNA extract prior to polymerase chain reaction (PCR) using Sigma′s REDExtract-N-Amp PCR Reaction Mix.

Legal Information

Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Zhang et al.
American journal of physiology. Renal physiology, 319(6), F1135-F1148 (2020-11-10)
Autosomal dominant polycystic kidney disease (ADPKD) is characterized by the presence of numerous fluid-filled cysts, extensive fibrosis, and the progressive decline in kidney function. Transforming growth factor-β1 (TGF-β1), an important mediator for renal fibrosis and chronic kidney disease, is overexpressed
Işın Çakır et al.
Nature metabolism, 4(1), 44-59 (2022-01-19)
The adipose tissue-derived hormone leptin can drive decreases in food intake while increasing energy expenditure. In diet-induced obesity, circulating leptin levels rise proportionally to adiposity. Despite this hyperleptinemia, rodents and humans with obesity maintain increased adiposity and are resistant to
A high-throughput functional genomics workflow based on CRISPR/Cas9-mediated targeted mutagenesis in zebrafish
Varshney GK, et al.
Nature Protocols, 11(12), 2357-2357 (2016)
Jun He et al.
Pharmacology, biochemistry, and behavior, 86(2), 327-333 (2006-12-16)
Adolescence is an important stage of brain development. Recent studies have indicated that neurogenesis in the brain occurs throughout life prompting comparisons of adolescent and adult neurogenesis. Since insulin-like growth factor 1 (IGF-1) has been implicated in promoting neurogenesis we
Yan Zhang et al.
American journal of physiology. Renal physiology, 319(6), F1135-F1148 (2020-11-10)
Autosomal dominant polycystic kidney disease (ADPKD) is characterized by the presence of numerous fluid-filled cysts, extensive fibrosis, and the progressive decline in kidney function. Transforming growth factor-β1 (TGF-β1), an important mediator for renal fibrosis and chronic kidney disease, is overexpressed

Protocols

SYBR® Green Extract-N-Amp™ Tissue PCR Kit enables rapid genomic DNA extraction and amplification from animal tissues.

SYBR® Green Extract-N-Amp™ Tissue PCR Kit enables rapid genomic DNA extraction and amplification from animal tissues.

SYBR® Green Extract-N-Amp™ Tissue PCR Kit enables rapid genomic DNA extraction and amplification from animal tissues.

SYBR® Green Extract-N-Amp™ Tissue PCR Kit enables rapid genomic DNA extraction and amplification from animal tissues.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service