Skip to Content
Merck
All Photos(2)

Key Documents

J4137

Sigma-Aldrich

JS-K

≥97%

Synonym(s):

O2-(2,4-Dinitrophenyl) 1-[(4-ethoxycarbonyl)piperazin-1-yl]diazen-1-ium-1,2-diolate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N6O8
CAS Number:
Molecular Weight:
384.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97%

storage condition

desiccated

solubility

DMSO: soluble 9.3 mg/mL
H2O: insoluble

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCOC(=O)N1CCN(CC1)\[N+]([O-])=N\Oc2ccc(cc2[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C13H16N6O8/c1-2-26-13(20)15-5-7-16(8-6-15)19(25)14-27-12-4-3-10(17(21)22)9-11(12)18(23)24/h3-4,9H,2,5-8H2,1H3/b19-14-

InChI key

DNJRNBYZLPKSHV-RGEXLXHISA-N

Biochem/physiol Actions

JS-K belongs to the class of diazeniumdiolate prodrug. It generates nitric oxide when metabolized by glutathione S-transferases (GST), which ensures optimal activity of JS-K. It has tumor growth inhibition action in human prostate cancer and leukemia.
Nitric oxide donor; antiproliferative.

Legal Information

Covered by US Patent No 6,610,660

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

JS-K, a GST-activated nitric oxide generator, induces DNA double-strand breaks, activates DNA damage response pathways, and induces apoptosis in vitro and in vivo in human multiple myeloma cells
Kiziltepe T, et al.s
Blood, 110(2), 709-718 (2007)
Bin Liu et al.
BMC cancer, 19(1), 645-645 (2019-07-03)
Ovarian cancer (OC) is the second most frequent gynecological cancer and is associated with a poor prognosis because OC progression is often asymptoma-tic and is detected at a late stage. There remains an urgent need for novel targeted therapies to
Rui Zhang et al.
Macromolecular bioscience, 16(1), 121-128 (2015-07-30)
There is a need for new treatment strategies of acute myeloid leukemia (AML). In this study, four different drugs, including cytarabine, daunorubicin, GDC-0980, and JS-K, were investigated in vitro for the two-drug combinations treatment of AML. The results revealed that
Zhengyun Liu et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 92, 989-997 (2017-06-14)
Hepatocellular carcinoma (HCC) is the most important cause of cancer-related death, and 85% of HCC is caused by chronic HBV infection, the prognosis of patients and the reduction of HBV DNA levels remain unsatisfactory. JS-K, a nitric oxide-releasing diazeniumdiolates, is
Ray Dong et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 88, 367-373 (2017-01-26)
JS-K is a novel anticancer nitric oxide (NO) prodrug effective against a variety of cancer cells, including the inhibition of AM-1 hepatoma cell growth in rats. To further evaluate anticancer effects of JS-K, human hepatoma Hep3B cells were treated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service