Skip to Content
Merck
All Photos(3)

Key Documents

M4139

Sigma-Aldrich

S-Methylglutathione

>98% (TLC), suitable for cell culture

Synonym(s):

L-γ-glutamyl-S-methyl-L-cysteinyl-glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19N3O6S
CAS Number:
Molecular Weight:
321.35
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

S-Methylglutathione,

Assay

>98% (TLC)

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

color

white

storage temp.

2-8°C

SMILES string

CSCC(NC(=O)CCC(N)C(O)=O)C(=O)NCC(O)=O

InChI

1S/C11H19N3O6S/c1-21-5-7(10(18)13-4-9(16)17)14-8(15)3-2-6(12)11(19)20/h6-7H,2-5,12H2,1H3,(H,13,18)(H,14,15)(H,16,17)(H,19,20)

InChI key

QTQDDTSVRVWHMO-UHFFFAOYSA-N

Biochem/physiol Actions

S-methylglutathione is a methionine containing peptide and glyoxylase inhibitor.

Substrates

Useful as a glyoxylase inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Michel Cailleret et al.
Circulation, 109(3), 406-411 (2004-01-21)
The negative effect of tumor necrosis factor-alpha (TNF-alpha) on heart contraction, which is mediated by sphingosine, is a major component in heart failure. Because the cellular level of glutathione may limit sphingosine production via the inhibition of the Mg-dependent neutral
Piotr Filipiak et al.
The journal of physical chemistry. B, 117(8), 2359-2368 (2013-01-26)
Nanosecond laser flash photolysis was used to generate sulfur radical cations of the thioether, S-methylglutathione (S-Me-Glu), via the one-electron oxidation of this thioether by triplet 4-carboxybenzophenone. The purpose of this investigation was to follow the neighboring group effects resulting from
J D Piganelli et al.
Diabetes, 47(8), 1212-1218 (1998-08-14)
IDDM results from the destruction of pancreatic beta-cells by autoreactive T-cells that appear to avoid deletion early in development, possibly due to improper interaction with antigen-presenting cells (APCs) resident in the thymus or periphery. In the nonobese diabetic (NOD) mouse
M F Phillips et al.
The Biochemical journal, 294 ( Pt 1), 57-62 (1993-08-15)
Mouse liver glutathione S-transferase YfYf (Pi class) reacts with [14C]ethacrynic acid to form a covalent adduct with a stoichiometry of 1 mol per mol of subunit. Proteolytic digestion of the enzyme-[14C]ethacrynic acid adduct with V8 protease produced an 11 kDa
João Costa Pessoa et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 7(3), 225-240 (2002-04-06)
The equilibria in the system V(IV)O(2+)-glutathione in aqueous solution were studied in the pH range 2-11 by a combination of pH-potentiometry and spectroscopy (EPR, visible absorption and circular dichroism). The results of the various methods are consistent and the equilibrium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service