Skip to Content
Merck
All Photos(1)

Key Documents

I2143

Sigma-Aldrich

Anti-Interleukin-6 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-IL-6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

neutralization: suitable using human IL-6 in a T1165 cell proliferation bioassay
radioimmunoassay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL6(3569)

General description

Anti-Interleukin 6 (IL-6) is produced in rabbit using purified human recombinant IL-6 produced in E. coli as the immunogen. Interleukin-6 is a multifunctional protein originally discovered in the media of cells stimulated with double stranded RNA. Interleukin-6 (IL-6) is a cytokine produced by activated T cells, endothelial cells, fibroblasts and activated monocytes or macrophages.

Specificity

The antibody will neutralize the biological activity of human IL-6 in cell culture. The antibody does not cross-react with human IL-1β, TNF-α or mouse IL-6.

Immunogen

Purified human recombinant IL-6 produced in E. coli.

Application

Anti-Interleukin-6 antibody can be used in western blotting and ELISA for studying human IL-6. It can also be used for dot blot immunoassay. Anti-Interleukin-6 antibody can be used for selective neutralization of human IL-6 bioactivity in cell culture. It can also be used in radioimmunoassay (RIA) (diluted 1:60,000).
It may also be used in immunoblotting.

Biochem/physiol Actions

Interleukin 6 (IL-6) acts upon a variety of cells, including fibroblasts, myeloid progenitor cells, T cells, B cells, and hepatocytes. Interleukin 6 (IL-6) also potentiates the proliferative effect of Interleukin 3 (IL-3) on multipotential hematopoietic progenitors.
Interleukin-6 (IL-6) is a cytokine produced by activated T cells, endothelial cells, fibroblasts and activated monocytes or macrophages. It plays a crucial role in regulating the acute phase response as well as responses occurs after injury and infection. It interacts with IL-2 and facilitates in the proliferation of T lymphocytes.

Physical form

Solution, 0.2 μm filtered, in 0.5 mL phosphate buffered saline, pH 7.4.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vuong Cat Khanh et al.
Stem cells and development, 29(21), 1382-1394 (2020-09-10)
Cancer metastasis is the leading cause of mortality among breast cancer patients. Type 2 diabetes mellitus (T2DM) has been suggested as a risk factor of breast cancer; however, whether or not T2DM is associated with breast tumor metastasis remains unclear.
Inflammation, obesity, stress and coronary heart disease: is interleukin-6 the link?
Yudkin J S, et al.
Atherosclerosis, 148(2), 209-214 (2000)
Debra J Romberger et al.
American journal of physiology. Lung cellular and molecular physiology, 311(1), L101-L110 (2016-05-18)
Agricultural dust exposure results in significant lung inflammation, and individuals working in concentrated animal feeding operations (CAFOs) are at risk for chronic airway inflammatory diseases. Exposure of bronchial epithelial cells to aqueous extracts of hog CAFO dusts (HDE) leads to
Interleukin-6 and chronic inflammation
Gabay C, et al.
Arthritis Research & Therapy, 8(2), S3-S3 (2006)
R P Nordan et al.
Science (New York, N.Y.), 233(4763), 566-569 (1986-08-01)
Plasmacytoma (PCT) cell lines dependent for proliferation and survival on a factor elaborated by the murine macrophage cell line, P388D1, were established in vitro. Adherent peritoneal cells induced by pristane produced 50-fold greater amounts of this activity in vitro than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service