Skip to Content
Merck
All Photos(1)

Key Documents

I2519

Sigma-Aldrich

Monoclonal Anti-Interleukin-8 antibody produced in mouse

clone 6217, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-CXCL8, Anti-IL-8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6217, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

capture ELISA: suitable
flow cytometry: suitable
immunocytochemistry: 25 μg/mL using using calcium ionophore- and PMA-stimulated human peripheral blood mononuclear cells
neutralization: suitable
western blot: 1-2 μg/mL

isotype

IgG1

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL8(3576)

General description

Interleukin-8 or CXCL8 (IL-8) is a proinflammatory chemokine that belongs to the C-X-C family. Monocytes and macrophages are the primary sources of IL-8 but it is also secreted by most nucleated cells. IL-8 binds to two G protein-coupled receptors, CXCR1 and CXCR2 and leads to the downstream activation of PI3K, MAPK, PKC, STAT3 and FAK pathways. IL-8 is responsible for the recruitment monocytes and neutrophils that results in a prolonged acute inflammation. The expression of IL-8 is influenced by inflammatory signals, chemical and environmental stress and hormones. Deregulation of IL-8 signaling has been observed in atherosclerosis, cardiovascular disease and solid tumors of bladder, prostate, ovary, glial cells and pancreas. Monoclonal Anti-Interleukin-8 antibody recognizes human IL-8. By immunoblotting, the antibody shows 100% cross reactivity with recombinant porcine IL-8, and no cross reactivity with recombinant rat CINC-2α.

Immunogen

purified, E. coli-derived, recombinant human IL-8.

Application

Monoclonal Anti-Interleukin-8 antibody may be used for neutralization assays; ND50 is 0.08-0.4 μg/ml. The antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. For ELISA a working concentration of 0.5 μg/ml may be used. Antibody concentration of 25 μg/ml may be used for immunocytochemistry using stimulated human peripheral blood mononuclear cells. For flow cytometry, 10 μL of 25 μg/ml stock antibody solution may be used for 1-5 X 105 cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Moira L Aitken et al.
Frontiers in physiology, 12, 653177-653177 (2021-05-11)
The role of inflammation in airway epithelial cells and its regulation are important in several respiratory diseases. When disease is present, the barrier between the pulmonary circulation and the airway epithelium is damaged, allowing serum proteins to enter the airways.
Daniel J Brat et al.
Neuro-oncology, 7(2), 122-133 (2005-04-16)
Interleukin-8 (IL-8, or CXCL8), which is a chemokine with a defining CXC amino acid motif that was initially characterized for its leukocyte chemotactic activity, is now known to possess tumorigenic and proangiogenic properties as well. In human gliomas, IL-8 is
The Interleukin-8 Pathway in Cancer
Waugh DJ and Wilson C
Clinical Cancer Research, 6735-6741 (2008)
C Knall et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(7), 3052-3057 (1997-04-01)
Chemoattractants and chemokines, such as interleukin 8 (IL-8), are defined by their ability to induce directed cell migration of responsive cells. The signal transduction pathway(s) leading to cell migration remain ill defined. We demonstrate that phosphatidylinositol-3-kinase (PI3K) activity, as determined
Pascal Wiegand et al.
Journal of the American Society for Mass Spectrometry, 31(1), 109-116 (2020-09-04)
The polypeptide chemokine Interleukin-8 (IL8) plays a crucial role in inflammatory processes in humans. IL8 is involved in chronic inflammatory lung diseases, rheumatoid arthritis, and cancer. Previous studies have shown that the interaction of IL8 with its natural receptors CXCR1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service