SRP3330
IL-6 from mouse
Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Synonym(s):
BSF-2, HPGF, IFN-?2, B cell differentiation factor (BCDF)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
mouse
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
potency
5x10^7 units/mg ED50
mol wt
21.7 kDa
packaging
pkg of 10 μg
impurities
endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... IL6(16193)
General description
Murine IL-6 is inactive on human cells, while both human and murine are equally active on murine cells. Recombinant murine IL-6 is a 21.7 kDa protein containing 188 amino acid residues.
Application
IL-6 from mouse has been used to study IL-6-mediated inhibition of mouse preadipocyte differentiation.
Biochem/physiol Actions
IL-6 is a pleiotropic cytokine that plays an important role in host defense by regulating immune and inflammatory responses. Produced by T cells, monocytes, fibroblasts, endothelial cells and keratinocytes, IL-6 has diverse biological functions. It stimulates B-cell differentiation and antibody production, synergizes with IL-3 in megakaryocyte development and platelet production, induces expression of hepatic acute-phase proteins, and regulates bone metabolism. IL-6 signals through the IL-6 receptor system that consists of two chains, IL-6R and gp130.
Physical form
Lyophilized from 0.1% TFA
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The biology of interleukin-6.
Kishimoto T
Blood, 74, 1-10 (1989)
Lichun Chen et al.
Foods (Basel, Switzerland), 11(20) (2023-07-11)
Dietary intervention is becoming more popular as a way to improve lipid metabolism and reduce the prevalence of diet-related chronic disorders. We evaluated the effects of several dietary oils on body weight, fat mass, liver weight, and tumor necrosis factor
Polyinosinic-polycytidylic acid inhibits the differentiation of mouse preadipocytes through pattern recognition receptor-mediated secretion of tumor necrosis factor-a.
Yu L
Immunology and Cell Biology, 94, 875-885 (2016)
T Kishimoto et al.
Science (New York, N.Y.), 258(5082), 593-597 (1992-10-23)
Many cytokines and cytokine receptors involved in the regulation of hematopoiesis, immune responses, and inflammation have been identified and characterized at the molecular level. Several characteristic features of cytokines, such as pleiotropy and redundancy, are now more clearly understood on
Lili Yu et al.
Immunology and cell biology, 94(9), 875-885 (2016-10-19)
Viral infections can disturb the functions of adipose tissues and thus result in metabolic diseases. Polyinosinic-polycytidylic acid (poly(I:C)), a synthetic analog of viral double-stranded RNA, induces innate antiviral responses by mimicking viral infection through the activation of pattern recognition receptors
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service