HS120245
Wide mouth wash bottle
white PE-LD bottle, capacity 500 mL
All Photos(1)
About This Item
UNSPSC Code:
41103209
NACRES:
NB.43
Recommended Products
material
polypropylene cap
white PE-LD bottle
feature
label no label
vented
with cap
packaging
pack of 6 ea
manufacturer/tradename
Heathrow Scientific HS120245
capacity
500 mL
Related Categories
General description
Color-coded tops for easy identification
The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
- Wide bottle mouth makes filling easier
- Color-coding for user, application or location identification
- Bottle is made from LDPE, with a PP closure
- 500 mL bottle size,
- Supplied in packs of 6
Choose from one of the most recent versions:
Certificates of Analysis (COA)