Z423114
Scienceware® safety labeled wash bottles
Ethanol
Synonym(s):
lab bottle, safety bottles, wash bottle
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103209
NACRES:
NB.17
Recommended Products
material
low-density polyethylene
feature
with cap
packaging
pack of 6 bottles
manufacturer/tradename
Bel-Art F11646-0639
capacity
500 mL
color
opaque
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Safety Labeled wash bottles with Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, for making your workplace a safer place.
Features and Benefits
- Easy-to-read printed bottles
- Wide mouth 53mm (2") bottle for easy refilling
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Polypropylene closures
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service