Skip to Content
Merck
All Photos(1)

Key Documents

GW23003

Sigma-Aldrich

Anti-CD19 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CD19

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-23003

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD19(930)

General description

CD19 is a 95kDa protein localized on the surface of nearly all B lymphocytes, and is considered to be the specific surface marker for B cells. This protein belongs to the immunoglobulin (Ig) superfamily with a cytoplasmic region of ~240 amino acids, and is involved in the regulation of B cell function. The protein is encoded by the 7.41kb cd19 gene, mapped on the short arm of human chromosome 16p11.2.
There are certain localized regions of strong net negative charge with nine tyrosine residues in the cytoplasmic region of CD19. These regions might regulate the interaction of CD19 with the proto-oncogene Vav, phosphatidylinositol-3 kinase (PI-3 kinase), and the Lyn, Lck, and Fyn protein tyrosine kinases.

Immunogen

B-lymphocyte antigen CD19.
Immunogen Sequence: GI # 23397636, sequence 101-291

Biochem/physiol Actions

CD19 expression is restricted to normal and malignant B-lineage cells. Therefore, it is considered to be a striking target for immunotherapies aimed at B-cell malignancies. CD19 helps B lymphocytes to initiate primary immune response even at low concentrations of antigen. The encoded protein plays a crucial role in initiating intrinsic B cell signaling thresholds by regulating both B cell receptor (BCR)-dependent and independent signaling. It also functions as a biomarker for B-cell development.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D T Fearon et al.
Annual review of immunology, 13, 127-149 (1995-01-01)
B lymphocytes must respond to low concentrations of antigen despite having low affinity antigen receptors during the primary immune response. CD19, a B cell-restricted membrane protein of the immunoglobulin superfamily that associates with the antigen receptor complex, may help the
James N Kochenderfer et al.
Blood, 119(12), 2709-2720 (2011-12-14)
We conducted a clinical trial to assess adoptive transfer of T cells genetically modified to express an anti-CD19 chimeric Ag receptor (CAR). Our clinical protocol consisted of chemotherapy followed by an infusion of anti-CD19-CAR-transduced T cells and a course of
A Pezzutto et al.
Journal of immunology (Baltimore, Md. : 1950), 138(9), 2793-2799 (1987-05-01)
The 95 Kd CD19 antigen is the broadest lineage specific surface marker for B cells: it is present on the surface of virtually all B lymphocytes, including early B progenitor cells. In this study we have evaluated the function of
L E Bradbury et al.
Journal of immunology (Baltimore, Md. : 1950), 149(9), 2841-2850 (1992-11-01)
CD19 is a member of the Ig superfamily expressed on the surface of B lymphocytes that may be involved in the regulation of B cell function. Immunoprecipitation studies with B cell lines solubilized by digitonin have shown CD19 to be
The CD19-CD21 complex regulates signal transduction thresholds governing humoral immunity and autoimmunity.
T F Tedder et al.
Immunity, 6(2), 107-118 (1997-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service