Skip to Content
Merck
All Photos(1)

Key Documents

GW21017

Sigma-Aldrich

Anti-Tenascin antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Cytotactin, Anti-Neuronectin, Anti-Tenascin C (hexabrachion)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21017

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNC(3371)

Immunogen

Immunogen Sequence: GI # 4504549, sequence 18-175
Recombinant tenascin C (hexabrachion)

Application

Anti-Tenascin antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200. It is also used for immunohistology.

Biochem/physiol Actions

Tenascin-C (TNC) is an extracellular matrix protein that is expressed at low levels in normal adult tissue but is highly expressed around many tumors including ovarian tumors. High levels are observed in many diseases like heart failure, thrombosis, atherosclerosis and cancer. It plays an indispensable role in tumor development. Expression of this gene is induced by growth factors as well as mechanical strain in fibroblast. It is up-regulated within glioma tissues and cultured glioma cell lines. TNC possesses a multi-modular structure with a variety of functional properties.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yucal Wang et al.
Oncology research, 20(11), 509-516 (2013-09-26)
Osteosarcoma (OS) is a kind of malignancy wherein the tumor cells form malignant bone-like or bone tissue. Tenascin-C (TN-C), an important extracellular matrix (ECM) protein, plays an indispensable role in tumor development. However, its regulatory factors, expression, and function in
D Tastekin et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(7), 6619-6625 (2014-04-04)
Tenascin-C (TNC) is a key molecule in tissue remodeling, and high levels are observed in many diseases, including heart failure, thrombosis, atherosclerosis, and cancer. High TNC expression by immunohistochemical analysis has been shown in invasive and metastasizing tissues from a
Tastekin Didem et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(7), 6777-6782 (2014-04-12)
Tenascin-C (TNC) is an extracellular matrix protein that is expressed at low levels in normal adult tissue but is highly expressed around many tumors including ovarian tumors. The objective of this study was to determine the clinical significance of the
Nicole Brösicke et al.
Cell and tissue research, 354(2), 409-430 (2013-08-22)
The extracellular matrix (ECM) protein tenascin-C (TN-C) is upregulated within glioma tissues and cultured glioma cell lines. TN-C possesses a multi-modular structure and a variety of functional properties have been reported for its domains. We describe five novel monoclonal antibodies
Daniel J Gibson et al.
Investigative ophthalmology & visual science, 54(7), 4776-4781 (2013-06-27)
To determine the topographical location and time course of development of corneal haze in a phototherapeutic keratectomy model using slit lamp examination, macrophotography, quantitative image analysis, and immunofluorescence staining of corneal sections. Rabbit corneas were ablated with an excimer laser

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service