Skip to Content
Merck
All Photos(1)

Key Documents

MAB1794

Sigma-Aldrich

Anti-Human B cells (CD19) Antibody, clone FMC63

clone FMC63, Chemicon®, from mouse

Synonym(s):

B-lymphocyte Surface Antigen B4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

FMC63, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunofluorescence: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD19(930)

General description

BIOCHEMISTRY:

p I = 7.0

Specificity

This specificity of this antibody is equivalent to that of CD19 monoclonal antibodies (Zola et al., 1988, 1989), based on comparative studies, antigen recognition and positive reaction with a CD19 transfected cell line. It detects a 96 kD single chain glycoprotein on the surface of human B lineage cells, including pre-B cells. B cells can be detected in peripheral blood and tissues. The antibody reacts with most B cell malignancies including chronic lymphocytic leukemia and most lymphomas, but not multiple myeloma.

Cell reactivity:

Normal:Stains all B lymphocytes in peripheral blood and spleen. Negative on granulocytes, monocytes, platelets, erythrocytes and T lymphocytes.

Clinical:

B cell chronic lymphocytic leukemia (B-CLL) +

Pro lymphocytic leukemia (PLL) +

Hairy cell leukemia (HCL) +

Common acute lymphoblastic leukemia (CALL) +

Pre-B acute lymphoblastic leukemia (pre-B-ALL) +

NULL-acute lymphoblastic leukemia (NULL-ALL) +

T common lymphocytic leukemia (T-CLL) -

Immunogen

Raji cells

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-Human B cells CD19 Antibody, clone FMC63 is validated for use in FC, IF for the detection of CD19.
This antibody is useful for enumerating normal B lymphocytes and typing B cell malignancies by flow cytometry and immunofluorescence.

SUGGESTED USAGE DILUTION

Flow cytometry and Indirect Immunofluorescence 1:25

Dilute with isotonic buffer. Use 50 μl of diluted antibody per 1 x 10E6 peripheral blood mononuclear cells (PBMC) in 100 μl buffer.

Linkage

Replaces: CBL141

Physical form

Format: Purified
Purified from mouse ascitic fluid. The antibody is supplied in 25 mM Tris-HCl buffer, 0.4 M NaCl, pH 8.0, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry.

Storage and Stability

Store at 2 to 8°C, for up to 6 months. For prolonged periods, store below -20°C in undiluted aliquots. AVOID REPEATED FREEZE/THAW CYCLES.

WARNING: The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabiana Zappala et al.
Science advances, 8(18), eabn4613-eabn4613 (2022-05-07)
Extensive antibody engineering and cloning is typically required to generate new bispecific antibodies. Made-to-order genes, advanced expression systems, and high-efficiency cloning can simplify and accelerate this process, but it still can take months before a functional product is realized. We
Ye Lin et al.
Frontiers in neurology, 13, 808322-808322 (2022-03-18)
Myasthenia gravis (MG) is an autoimmune disease mediated by B cells secreting autoantibodies. Regulatory B (Breg) cells confirmed to have an immunosuppressive function play an important role in many autoimmune diseases. However, what about the changes in Breg cells in
Stephanie N Shishido et al.
Journal for immunotherapy of cancer, 12(2) (2024-02-14)
Chimeric antigen receptor (CAR)-T cells are approved for use in the treatment of hematological malignancies. Axicabtagene ciloleucel (YESCARTA) and brexucabtagene autoleucel (TECARTUS) genetically modified autologous T cells expressing an anti-CD19 scFv based on the FMC63 clone have shown impressive response
Christine Ambrose et al.
PloS one, 16(3), e0247701-e0247701 (2021-03-19)
Successful CAR T cell therapy for the treatment of solid tumors requires exemplary CAR T cell expansion, persistence and fitness, and the ability to target tumor antigens safely. Here we address this constellation of critical attributes for successful cellular therapy
Xing Du et al.
Cancer research, 68(15), 6300-6305 (2008-08-05)
B-cell malignancies routinely express surface antigens CD19 and CD22. Immunotoxins against both antigens have been evaluated, and the immunotoxins targeting CD22 are more active. To understand this disparity in cytotoxicity and guide the screening of therapeutic targets, we compared two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service