Skip to Content
Merck
All Photos(3)

Key Documents

G9793

Sigma-Aldrich

N-Glycolylneuraminic acid

≥95% (HPLC), semisynthetic

Synonym(s):

Neu5Glc, NeuNGl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO10
CAS Number:
Molecular Weight:
325.27
Beilstein:
1716828
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

semisynthetic

Quality Level

Assay

≥95% (HPLC)

form

powder

technique(s)

LC/MS: suitable

impurities

water (Karl Fischer)

color

white

solubility

water: soluble 20 mg/mL

suitability

suitable for LC-MS

application(s)

metabolomics

storage temp.

−20°C

SMILES string

[H][C@]1(O[C@@](O)(C[C@H](O)[C@H]1NC(=O)CO)C(O)=O)[C@H](O)[C@H](O)CO

InChI

1S/C11H19NO10/c13-2-5(16)8(18)9-7(12-6(17)3-14)4(15)1-11(21,22-9)10(19)20/h4-5,7-9,13-16,18,21H,1-3H2,(H,12,17)(H,19,20)/t4-,5+,7+,8+,9+,11-/m0/s1

InChI key

FDJKUWYYUZCUJX-AJKRCSPLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Glycolylneuraminic acid (Neu5Gc) is a nonhuman sialic acid molecule synthesized in pigs but not in humans, that has a role as a mammalian metabolite and an antigen. This hydroxylated form of sialic acid, specifically N-acetylneuraminic acid, features a glycolyl substituent on nitrogen and a beta-configuration at the anomeric center. The biosynthesis of N-glycolylneuraminic acid involves the enzymatic activity of cytidine monophosphate-N-acetylneuraminate (CMP-Neu5Ac) hydroxylase.
In humans, the absence of endogenous production results from a gene mutation affecting CMP-Neu5Ac hydroxylase, the enzyme responsible for converting N-acetylneuraminic acid into Neu5Gc. However, Neu5Gc can accumulate in human cells through external ingestion from dietary sources like red meat and dairy products. N-Glycolylneuraminic acid is a versatile compound that finds application in cell biology, metabolomics and biochemical research

Application

N-Glycolylneuraminic acid has been used:
  • as a sugar in microtiter biofilm methodologic approach for the enhancement of biofilm formation
  • as a standard for the determination of sialic acids in the nervous system of silkworm and to find the variations of sialic acids among different developmental stages.
  • as a standard in the high-performance liquid chromatography (HPLC) analyses to detect the molecular species of sialic acid (Sia) species using 1,2-diamino-4,5-methylenedioxy-benzene (DMB) as a fluorogenic compound

Biochem/physiol Actions

N-Glycolylneuraminic acid (Neu5Gc) functions as an important tool for profiling anti-Neu5Gc antibodies and sialic acid-binding proteins.

Features and Benefits

  • Ideal for Metabolomics, Biochemical and Cell Biology research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

comparable product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anoopjit Singh Kooner et al.
Frontiers in immunology, 10, 2004-2004 (2019-09-27)
Sialic acids constitute a family of negatively charged structurally diverse monosaccharides that are commonly presented on the termini of glycans in higher animals and some microorganisms. In addition to N-acetylneuraminic acid (Neu5Ac), N-glycolyl neuraminic acid (Neu5Gc) is among the most
Chao-Tan Guo et al.
Glycobiology, 17(7), 713-724 (2007-03-29)
The receptor specificity of influenza viruses is one factor that allows avian influenza viruses to cross the species barrier. The recent transmissions of avian H5N1 and H9N2 influenza viruses from chickens and/or quails to humans indicate that avian influenza viruses
Nobuyoshi Hayashi et al.
Cancer science, 104(1), 43-47 (2012-09-26)
Gangliosides are glycosphingolipids found on the cell surface. They act as recognition molecules or signal modulators and regulate cell proliferation and differentiation. N-glycolylneuraminic acid (NeuGc)-containing gangliosides have been detected in some neoplasms in humans, although they are usually absent in
Dzung H Nguyen et al.
Journal of immunology (Baltimore, Md. : 1950), 175(1), 228-236 (2005-06-24)
Humans are genetically incapable of producing the mammalian sialic acid N-glycolylneuraminic acid (Neu5Gc), due to an inactivating mutation in the enzyme synthesizing it. Despite this, human cells and tissues appear capable of metabolically incorporating Neu5Gc from exogenous sources, including dietary
Xing Yu et al.
Journal of virology, 86(24), 13456-13466 (2012-10-05)
The rotavirus spike protein domain VP8* is essential for recognition of cell surface carbohydrate receptors, notably those incorporating N-acylneuraminic acids (members of the sialic acid family). N-Acetylneuraminic acids occur naturally in both animals and humans, whereas N-glycolylneuraminic acids are acquired

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service