Skip to Content
Merck
All Photos(1)

Key Documents

G3796

Sigma-Aldrich

Granisetron hydrochloride

≥98% (HPLC), solid

Synonym(s):

1-Methyl-N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N4O· HCl
CAS Number:
Molecular Weight:
348.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to off-white

mp

284.0-290.0 °C

solubility

H2O: >10 mg/mL

originator

Roche

storage temp.

room temp

SMILES string

Cl.CN1[C@H]2CCC[C@@H]1C[C@@H](C2)NC(=O)c3nn(C)c4ccccc34

InChI

1S/C18H24N4O.ClH/c1-21-13-6-5-7-14(21)11-12(10-13)19-18(23)17-15-8-3-4-9-16(15)22(2)20-17;/h3-4,8-9,12-14H,5-7,10-11H2,1-2H3,(H,19,23);1H/t12-,13+,14-;

InChI key

QYZRTBKYBJRGJB-WQTKJZBYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Granisetron hydrochloride helps to block nausea/emesis and abdominal pain in irritable bowel syndrome, that is stimulated by chemotherapy.[1]
Granisetron hydrochloride is a serotonin 5-HT3 receptor antagonist and antiemetic.

Features and Benefits

This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alterations of colonic sensitivity and gastric dysmotility after acute cisplatin and granisetron
Martin R, et al.
Neurogastroenterology and Motility, e13499-e13499 (2018)
Hea-Jeong Doh et al.
Colloids and surfaces. B, Biointerfaces, 101, 475-480 (2012-09-27)
A new lipid nanoemulsion (LNE) system containing granisetron (GRN) was developed and its in vitro permeation-enhancing effect was evaluated using Caco-2 cell monolayers. Particle size, polydispersity index (PI) and stability of the prepared GRN-loaded LNE systems were also characterized. The
Caroline Sévoz-Couche et al.
The Journal of physiology, 591(7), 1871-1887 (2013-01-09)
Anxiety disorders in humans reduce both the heart rate variability (HRV) and the sensitivity of the cardiac baroreflex (BRS). Both may contribute to sudden death. To elucidate the mechanisms underlying these alterations, male rats were subjected to social defeat sessions
Mohammad-Reza Khodaie-Ardakani et al.
Journal of psychiatric research, 47(4), 472-478 (2013-02-05)
Some 5-HT3 antagonists such as ondansetron have shown beneficial effects on negative symptoms of patients with schizophrenia. We aimed to evaluate the efficacy of granisetron (another 5-HT3 antagonist) add-on therapy in the treatment of negative symptoms of patients with stable
Divya Kesters et al.
EMBO reports, 14(1), 49-56 (2012-12-01)
The 5-HT(3) receptor is a pentameric serotonin-gated ion channel, which mediates rapid excitatory neurotransmission and is the target of a therapeutically important class of anti-emetic drugs, such as granisetron. We report crystal structures of a binding protein engineered to recognize

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service