Skip to Content
Merck
All Photos(1)

Key Documents

F100

Sigma-Aldrich

Fluspirilene

Synonym(s):

8-[4,4-bis(p-Fluorophenyl)butyl]-1-phenyl-1,3,8-triazino[4.5]decan-4-one, R 6218

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H31F2N3O
CAS Number:
Molecular Weight:
475.57
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: ≥20 mg/mL
H2O: insoluble
ethanol: soluble

originator

Johnson & Johnson

storage temp.

room temp

SMILES string

Fc1ccc(cc1)C(CCCN2CCC3(CC2)N(CNC3=O)c4ccccc4)c5ccc(F)cc5

InChI

1S/C29H31F2N3O/c30-24-12-8-22(9-13-24)27(23-10-14-25(31)15-11-23)7-4-18-33-19-16-29(17-20-33)28(35)32-21-34(29)26-5-2-1-3-6-26/h1-3,5-6,8-15,27H,4,7,16-21H2,(H,32,35)

InChI key

QOYHHIBFXOOADH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluspirilene has been used as a neuroleptic drug to study its effects on human ether-a-go-go related gene (HERG).
Fluspirilene has been used to study its cytotoxicity effect on malignant glioma.

Biochem/physiol Actions

Fluspirilene has a potential to inhibit the activity of cyclin-dependent kinase 2 (CDK2). It is an effective anti-cancer agent used for treating human hepatocellular carcinoma.
Fluspirilene is a dopamine receptor antagonist. It also acts as a calcium channel blocker. Fluspirilene is an antipsychotic agent and exhibits neuoleptic properties. It exhibits therapeutic effects against glioblastoma and Schizophrenia.

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Zalejski et al.
Plant physiology, 141(4), 1555-1562 (2006-06-13)
Diacylglycerol pyrophosphate (DGPP) was recently shown to be a possible intermediate in abscisic acid (ABA) signaling. In this study, reverse transcription-PCR of ABA up-regulated genes was used to evaluate the ability of DGPP to trigger gene expression in Arabidopsis (Arabidopsis
Su-Jane Wang
Synapse (New York, N.Y.), 44(1), 36-41 (2002-02-14)
Fluspirilene, a neuroleptic drug which is used clinically to treat schizophrenic patients, is a dopamine D2 receptor antagonist. Besides its well-known actions on the dopamine receptors, fluspirilene also displays calcium channel-blocking activity. The aim of this study was to investigate
Depot fluspirilene for schizophrenia
Abhijnhan A, et al.
Cochrane database of systematic reviews (Online), 8(1), 111728-111728 (2007)
D W Sah et al.
Molecular pharmacology, 45(1), 84-92 (1994-01-01)
P-type Ca2+ channels in cerebellar Purkinje neurons and N-type Ca2+ channels in sympathetic neurons were found to be inhibited by D2 dopamine receptor antagonists with diverse structures, including phenothiazines (chlorpromazine and thioridazine), diphenylbutylpiperidines (fluspirilene and pimozide), butyrophenones (haloperidol and spiperone)
Identification of antipsychotic drug fluspirilene as a potential anti-glioma stem cell drug
Dong Y, et al.
Oncotarget, 8(67), 111728-111728 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service