Skip to Content
Merck
All Photos(5)

Key Documents

H9523

Sigma-Aldrich

Serotonin hydrochloride

≥98%, powder, neurotransmitter

Synonym(s):

3-(2-Aminoethyl)-5-hydroxyindole hydrochloride, 5-HT, 5-Hydroxytryptamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O · HCl
CAS Number:
Molecular Weight:
212.68
Beilstein:
3570963
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Serotonin hydrochloride, powder

Assay

≥98%

Quality Level

form

powder

color

white to off-white

mp

149-154 °C (lit.)

solubility

H2O: 17 mg/mL
0.1 M HCl: 22 mg/mL
ethanol: 5.3 mg/mL

storage temp.

2-8°C

SMILES string

Cl[H].NCCc1c[nH]c2ccc(O)cc12

InChI

1S/C10H12N2O.ClH/c11-4-3-7-6-12-10-2-1-8(13)5-9(7)10;/h1-2,5-6,12-13H,3-4,11H2;1H

InChI key

MDIGAZPGKJFIAH-UHFFFAOYSA-N

Gene Information

human ... HTR3A(3359)

Looking for similar products? Visit Product Comparison Guide

General description

Serotonin is a ubiquitous molecule found in all animals. It shows diverse effects in the central and peripheral nervous system (CAN/PNS). Serotonin acts as a hormone and as a mitogen. It is produced in the axon terminal present in the brain. Action potential induces the release of serotonin that diffuses across the synapse to turn on the post-synaptic receptors. Serotonin plays a crucialrole in development, respiration, thermoregulation, endocrine, cardiovascular, and gastrointestinal functions. It also plays a role in sensory perceptional behaviors such as the sleep-wake cycle, sex, mood, memory, aggression, pain sensitivity, learning, and cognition. Serotonin impacts several psychiatric disorders such as schizophrenia, anorexia nervosa, depression, and a range of anxiety disorders such as obsessive-compulsive disorder, panic attacks, and social phobia. Variations in serotonin levels are associated withimpulse-related disorders or personalities like substance abuse, gambling, attention deficit disorder, obsessive-control, and aggression.

Application

Serotonin hydrochloride hasbeen used: to study the mechanisms of 5-hydroxytryptamine receptor antagonistsregulate fibrosis in liver spheroids, to study its effects on theextracellular-related-kinase (ERK) phosphorylation in human embryonic kidneycells, in transwell assay, wound healing assay and tumor allograft assay incolon carcinoma cell line

Biochem/physiol Actions

Neurotransmitter.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Solutions should be freshly prepared.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soaleha Shams et al.
Behavioral neuroscience, 131(6), 492-504 (2017-12-01)
Social isolation can be used to study behavioral, neural, and hormonal mechanisms that regulate interactions in social animals. Although isolation effects have been reported in social mammals and various fish species, systematic studies with isolated zebrafish are rare. Here, the
Serotonin Receptors
Nichols David E and Nichols Charles D.
Chemical Reviews (2008)
Vinay Kumar Sharma et al.
Research square (2023-02-16)
5-Hydroxy tryptamine receptor 1E (5-HTR1E) is reported to activate cAMP and ERK pathways via its ligands and binding partners, but the detailed mechanism underlying the serotonin induced 5-HTR1E signaling is still not known. In the present study, we determined the
Sara Redenšek Trampuž et al.
Scientific reports, 14(1), 1396-1396 (2024-01-17)
Non-alcoholic steatohepatitis (NASH) is a major health problem leading to liver fibrosis and hepatocellular carcinoma, among other diseases, and for which there is still no approved drug treatment. Previous studies in animal models and in LX-2 cells have indicated a
I Lucki
Biological psychiatry, 44(3), 151-162 (1998-08-07)
The diverse array of behavioral effects of serotonin form the basis for understanding its potential role as an etiological marker in psychiatric disorders and for the successful pharmacologic intervention of drugs regulating serotonin neurotransmission in behavior. General theories of the

Articles

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service