Skip to Content
Merck
All Photos(1)

Key Documents

F2266

Sigma-Aldrich

Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–FITC antibody produced in sheep

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

sheep

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:128

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgGs are glycoprotein antibodies that regulate immunological defense mechanisms such as phagocytosis and inflammatory responses . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice . Anti-Mouse IgG (whole molecule) F(ab′)2 fragment-FITC antibodies are specific for mouse IgG subclasses G1, G2a, G2b, and G3 and do not react with serum proteins in rat.

Immunogen

Mouse IgG

Application

Anti-Mouse IgG (whole molecule) F(ab′)2 fragment-FITC antibody is suitable for use in flow cytometry, immunohistochemistry, indirect immunofluorescence, and immunohistochemistry .
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Other Notes

Antibody adsorbed with rat serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with rat samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Mechaly et al.
Applied and environmental microbiology, 74(3), 818-822 (2007-10-30)
A single-chain Fv (scFv) antibody was developed and applied for efficient and specific detection of Bacillus anthracis spores. The antibody was isolated from a phage display library prepared from spleens of mice immunized with a water-soluble extract of the outer
C M Newman et al.
Arteriosclerosis, thrombosis, and vascular biology, 15(11), 2010-2018 (1995-11-01)
Osteopontin (OP) is a secreted glycoprotein that contains the Arg-Gly-Asp (RGD) cell-binding sequence that binds calcium and is chemotactic and adhesive for rat vascular smooth muscle cells (VSMCs). OP gene expression is upregulated in cultured rat VSMCs in vitro and
Y Yan et al.
Journal of immunology (Baltimore, Md. : 1950), 166(8), 5258-5264 (2001-04-06)
Donor leukocytes play a dual role in rejection and acceptance of transplanted organs. They provide the major stimulus for rejection, and their removal from the transplanted organ prolongs its survival. Paradoxically, administration of donor leukocytes also prolongs allograft survival provided
Hisashi Ueta et al.
Hepatology (Baltimore, Md.), 47(4), 1352-1362 (2008-01-29)
Donor dendritic cell (DC) migration and allosensitization in host secondary lymphoid organs after liver transplantation are ill defined. We used rat models to investigate graft-derived cells and intrahost allosensitization. Liver transplantation induced diffuse blood-borne migration of donor major histocompatibility class
Lech Kiedrowski et al.
Journal of neurochemistry, 90(1), 117-128 (2004-06-17)
Inhibition of Na(+),K(+)-ATPase during NMDA applications greatly increased NMDA-induced excitotoxicity in primary cultures of forebrain neurons (FNs), but not in cerebellar granule cells (CGCs). Because Na(+),K(+)-ATPase inhibition promotes reversal of plasmalemmal Na(+)/Ca(2+) exchangers, we compared the activities of reversed K(+)-independent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service