Skip to Content
Merck
All Photos(1)

Key Documents

F9006

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (whole molecule)–Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

FITC conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:64

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibody consists of four subclasses : IgG1, IgG2, IgG3 and IgG4. The IgG structure possesses four polypeptide chains containing two identical γ heavy (H) chains and two identical κ or λ light (L) chains of 50 kDa and 25 kDa respectively. The chains are interlinked with a disulfide bond.
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections
Anti-Mouse IgG (whole molecule)-FITC antibody is specific for mouse IgG. Goat anti-mouse IgG is conjugated to Fluorescein Isothiocyanate (FITC), Isomer I. Following conjugation, unbound FITC is removed by extensive dialysis.

Immunogen

Purified mouse IgG

Application

Anti-Mouse IgG (whole molecule)−FITC antibody produced in goat has been used in flow cytometry.
The antibody may be used for immunofluorescent staining of mouse spleen cells at a working dilution of 1:64. A working antibody dilution of 1:40 was used to stain the conidia of Botrytis cinerea for immunofluorescence. The antibody was also used for immunofluorescence of primary luteal cell cultures incubated with primary antibody against CD45 and rhizoids of Chara globularis incubated with anti-human spectrin antibody.

Biochem/physiol Actions

IgG (immunoglobulin G) antibody has its function similar to IgM antibody in complement system activation. IgG participates in hypersensitivity type II and type III. It helps in opsonization, complement fixation and antibody-dependent cell-mediated cytotoxicity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a monoclonal antibody-based immunodetection assay for Botrytis cinerea
Bossi R and Dewey FM
Plant Pathology, 41, 472-482 (1992)
Veronica Lanza Cariccio et al.
mAbs, 8(4), 741-750 (2016-03-11)
There is a strong need for rapid and reliable epitope mapping methods that can keep pace with the isolation of increasingly larger numbers of mAbs. We describe here the identification of a conformational epitope using Phage-based Representation OF ImmunoLigand Epitope
Specific IgG for cat allergens in patients with allergic conjunctivitis
The Laboratory Rat, 35(4), 575-586 (2005)
N C Crespo et al.
Cell death and differentiation, 9(7), 702-709 (2002-06-12)
Recently, we have shown that the farnesyltransferase inhibitor FTI-2153 induces accumulation of two human lung cancer cell lines in mitosis by inhibiting bipolar spindle formation during prometaphase. Here we investigate whether this mitotic arrest depends on transformation, Ras and/or p53
M Braun
Plant physiology, 125(4), 1611-1619 (2001-04-12)
Spectrin-like epitopes were immunochemically detected and immunofluorescently localized in gravitropically tip-growing rhizoids and protonemata of characean algae. Antiserum against spectrin from chicken erythrocytes showed cross-reactivity with rhizoid proteins at molecular masses of about 170 and 195 kD. Confocal microscopy revealed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service