Skip to Content
Merck
All Photos(2)

Key Documents

F9137

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−FITC antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Rabbit Anti-Mouse IgG (whole molecule)−Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

FITC conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

direct immunofluorescence: 1:100
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibodies are the smallest immunoglobulins found in all body fluids and can fight against bacterial and viral infections. It is the only type of antibody that can cross the placenta in pregnant woman and give immunity to the foetus. Anti-mouse IgG (whole molecule)-FITC antibody can be used as a secondary antibody in FACS analysis to detect cell surface expression of cd59b and cd59a in cloned CHO cells. Rabbit anti-mouse IgG (whole molecule)-FITC antibody reacts specifically with mouse IgG, IgA and IgM.
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. Anti-Mouse IgG (whole molecule)-FITC antibody reacts with all mouse IgG subclasses and IgM. Purified rabbit anti-Mouse IgG is conjugated to FITC in an alkaline reaction.

Specificity

Specificity for mouse IgG is determined by immunoelectrophoresis (IEP) prior to conjugation using normal mouse serum and mouse IgG. Product reacts with mouse IgA and IgM.

Immunogen

IgG from pooled normal mouse serum

Application

Anti-Mouse IgG (whole molecule)-FITC antibody may be used for immunofluorescence at a working antibody dilution of 1:100 on human tonsil sections using mouse monoclonal anti-human IgG as primary antibody. For immunofluorescence of human peripheral blood lymphocytes, a minimum dilution of 1:100 is recommended. For immunohistochemistry applications, the working antibody dilutions range from 1:100 to 1:1000 in various studies.
Anti-mouse IgG (whole molecule)-FITC antibody (diluted 1:1000) can be used as secondary antibody in immunohistochemistry.
Anti-Mouse IgG (whole molecule)-FITC antibody has been used in immunofluorescence and cell proliferation assay.

Biochem/physiol Actions

Immunoglobulin G (IgG) mainly helps in immune defense. It participates in hypersensitivity type II and type III. It also helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at 2-8 °C for a maximum of one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nakako Izumi Nakajima et al.
PloS one, 8(8), e70107-e70107 (2013-08-24)
Heavy particle irradiation produces complex DNA double strand breaks (DSBs) which can arise from primary ionisation events within the particle trajectory. Additionally, secondary electrons, termed delta-electrons, which have a range of distributions can create low linear energy transfer (LET) damage
Federica Zacchini et al.
Cellular reprogramming, 13(6), 495-502 (2011-11-03)
The production of androgenetic embryos in large animals is a complex procedure. Androgenetic embryos have been produced so far only in cattle and sheep using pronuclear transfer (PT) between zygotes derived from in vitro fertilization (IVF) of previously enucleated oocytes.
Tao Huang et al.
Experimental and therapeutic medicine, 14(4), 2831-2838 (2017-10-03)
In view of the compelling anti-diabetic effects of gastric bypass surgery (GBS) in the treatment of morbid obesity, it is important to clarify its enhancing effect on pancreatic islets, which is closely linked with diabetes remission in obese patients, as
Jeng-Chun Mei et al.
Tissue engineering. Part A, 20(21-22), 2931-2941 (2014-05-24)
To repair damaged cardiac tissue, the important principle of in vitro cell culture is to mimic the in vivo cell growth environment. Thus, micro-sized cells are more suitably cultured in three-dimensional (3D) than in two-dimensional (2D) microenvironments (ex: culture dish).
Regulation of Claspin degradation by the ubiquitin-proteosome pathway during the cell cycle and in response to ATR-dependent checkpoint activation.
Bennett LN and Clarke PR
Febs Letters, 580(17), 4176-4181 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service