EP0030501306
Eppendorf® Deepwell plates, standard, 96 wells
white plate, conical bottom colorless wells, capacity 2000 μL, pkg of 20 ea
About This Item
Recommended Products
material
conical bottom colorless wells
white plate
sterility
sterile; autoclaved
packaging
pkg of 20 ea
manufacturer/tradename
Eppendorf® 0030501306
availability
available only in (India)
technique(s)
PCR: suitable
capacity
2000 μL
size
96 wells
working volume
2 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Eppendorf® Deepwell plates, standard, 96 wells are high performance plates of capacity 2000 μL, manufactured from pure polypropylene (PP).
- These plates offer innovative features such as more dependability, more efficiency and more stability.
- These plates are available in white border and for easy identification of the wells, a high contrast alphanumeric label is laser-engraved in the border.
- This OptiTrack® matrix allows you to find your sample up to 30% more quickly while reducing pipetting errors.
- RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties.
- Minimal remaining volume and high uniformity from well to well.
- Raised well rims and smooth surface for reliable closing, even with heat sealing.
- Easy and reliable stacking.
- g-Safe®: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality.
Application
- Sample storage and preparation Bacteria and yeast cultivation
- High-throughput DNA and RNA isolation
- Storage of cDNA or genomic libraries
- Storage of siRNA or oligonucleotide libraries
- Protein analysis Storage of valuable protein and DNA samples in Eppendorf LoBind Plates
- High-throughput plasmid purification Creation of dilution series
Features and Benefits
- OptiTrack matrix: up to 30 % faster sample detection and fewer pipetting errors thanks to high-contrast alphanumeric labeling
- RecoverMax well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
- g-Safe : exceptional centrifugation resistance up to 6,000 × g for faster protocols and improved sample quality
- Raised corrugated edges and smooth surface for reliable sealing, even during heat sealing
- Easily and reliably stackable
- Minimal residual volume and high homogeneity from well to well
- High-quality polypropylene for high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without mold release agents, plasticizers and biocides – substances that can negatively affect bioassays
- Autoclavable (121 C, 20 min)
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service