Skip to Content
Merck
All Photos(1)

Key Documents

BR701354

BRAND® 96-well deep well plate, stackable

volume 2.2 mL, polypropylene

Synonym(s):

Deep-well Plates, Sample Storage

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

polypropylene

description

alphanumeric identification system

sterility

non-sterile

feature

skirt

packaging

pack of 24 ea

manufacturer/tradename

BRAND 701354

size

96 well

volume

2.2 mL

working volume

2.2 mL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

BRAND® 96-deep-well plate is a versatile laboratory tool designed for various applications. Made of polypropylene (PP), this deep-well plate features a U-bottom (square) shape and can accommodate 2.2 mL.

Application

BRAND® 96 deep well plates are suitable for use in:
  • Cell culture and tissue culture
  • High-Throughput Screening (HTS)
  • Tests requiring a mother plate
  • Serial dilutions
  • Reagent transfer and sample storage down to -80 °C (PP)

Features and Benefits

  • BRAND® 96-well deep well plates are manufactured according to ANSI/SLAS 1 and 4
  • Stackable for easy storage
  • The ′U′ bottom ensures optimal sample mixture and collection

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Accurate steroid analysis in blood serum using Supel™ Swift HLB DPX Tips; UTAK and NIST standards.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service