EP0030501209
Eppendorf® Deepwell plates, standard, 96 wells
white plate, conical bottom colorless wells, capacity 1000 μL, pkg of 20 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
NACRES:
NB.15
Recommended Products
material
conical bottom colorless wells
white plate
sterility
non-sterile
packaging
pkg of 20 ea
manufacturer/tradename
Eppendorf® 0030501209
availability
available only in (India)
technique(s)
PCR: suitable
capacity
1000 μL
size
96 wells
working volume
1 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Eppendorf® Deepwell Plates 96 are high-performance plates for all manual and automated applications − from sample storage at -86 ºC to DNA denaturation at 100 ºC.
- Manufactured from pure polypropylene (PP), these plates offer innovative features: More dependability. More efficiency. More stability.
- The plates are available with a white, yellow, green, or blue border. For easy identification of wells, a high contrast alphanumeric label is laser-engraved in the border.
- This OptiTrack® matrix allows you to find your sample up to 30% more quickly while reducing pipetting errors.
For greater reliability, stability and efficiency, choose Eppendorf Deepwell 96 well Plates. Available with white, yellow, green or blue frames, these deepwell plates offer high performance for all applications - both manual and automated. Thanks to OptiTrack matrix, a high-contrast alphanumeric label laser-engraved into the frame, you can find your sample up to 30% faster and reduce pipetting errors. For every application, from sample storage at 86 C to DNA denaturation at 100 C, rely on Eppendorf Deepwell Plates and set new standards in your laboratory.
Application
- Sample storage and preparation.
- Bacteria and yeast cultivation.
- DNA and RNA isolation with high throughput.
- Storage of cDNA or genomic banks.
- Storage of siRNA or oligonucleotide libraries.
- Protein analysis.
- Storage of valuable protein and DNA samples in Eppendorf LoBind plates.
- Plasmid purification with high throughput.
- Creating dilution series.
- Sample storage and preparation Bacteria and yeast cultivation
- High-throughput DNA and RNA isolation
- Storage of cDNA or genomic libraries
- Storage of siRNA or oligonucleotide libraries
- Protein analysis Storage of valuable protein and DNA samples in Eppendorf LoBind Plates
- High-throughput plasmid purification Creation of dilution series
Features and Benefits
- OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling.
- RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties.
- Minimal remaining volume and high uniformity from well to well.
- Raised well rims and smooth surface for reliable closing, even with heat sealing.
- Easy and reliable stacking.
- g-Safe®: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality.
- High-quality polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes.
- Manufactured without slip agents, plasticizers, and biocides − Materials which could have a negative effect on bioassays.
- Autoclavable (121 °C, 20 min).
- OptiTrack matrix: up to 30 % faster sample detection and fewer pipetting errors thanks to high-contrast alphanumeric labeling
- RecoverMax well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
- g-Safe : exceptional centrifugation resistance up to 6,000 × g for faster protocols and improved sample quality
- Raised corrugated edges and smooth surface for reliable sealing, even during heat sealing
- Easily and reliably stackable
- Minimal residual volume and high homogeneity from well to well
- High-quality polypropylene for high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without mold release agents, plasticizers and biocides – substances that can negatively affect bioassays
- Autoclavable (121 C, 20 min)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service