Skip to Content
Merck
All Photos(1)

Key Documents

E8515

Sigma-Aldrich

β-Estradiol 3-benzoate

≥97%

Synonym(s):

1,3,5(10)-Estratriene-3,17β-diol 3-benzoate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 3-benzoate, 3-Benzoyloxy-17β-estrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28O3
CAS Number:
Molecular Weight:
376.49
Beilstein:
3107526
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥97%
Pricing and availability is not currently available.

Quality Level

Assay

≥97%

mp

191-198 °C (lit.)

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)CC[C@@]3([H])[C@]1([H])CCc4cc(OC(=O)c5ccccc5)ccc24

InChI

1S/C25H28O3/c1-25-14-13-20-19-10-8-18(28-24(27)16-5-3-2-4-6-16)15-17(19)7-9-21(20)22(25)11-12-23(25)26/h2-6,8,10,15,20-23,26H,7,9,11-14H2,1H3/t20-,21-,22+,23+,25+/m1/s1

InChI key

UYIFTLBWAOGQBI-BZDYCCQFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-Estradiol 3-benzoate has been used:to mimic estradiol in rat during the oestrous cycle in rats.[1] It has also been used along with progesterone for the induction of sexual receptivity in female rat.[2]
Used in a study of general steroid metabolism and clearing, as groundwork for anabolic steroid screening in athletes.

Biochem/physiol Actions

β-Estradiol 3-benzoate is a synthetic estrogen. It favors breast cancer progression.[3] It is an endocrine disrupting chemicals. It interacts with endocrine system and cause harmful effect on reproduction in animals and humans. It binds to estrogen receptors and favors tumor metastatis in breast, prostate and bone cells.[4]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical sensing of estradiol benzoate using hydroxyapatite with three-dimensional channel frameworks
Wen Y, et al.
Analytical Methods : Advancing Methods and Applications, 9(40), 5868-5872 (2017)
Sexual behavior increases cell proliferation in the rostral migratory stream and promotes the differentiation of the new cells into neurons in the accessory olfactory bulb of female rats
Corona R, et al.
Frontiers in Neuroscience, 10, 48-48 (2016)
M A Mohr et al.
Hormones and behavior, 111, 110-113 (2018-12-16)
In females, a hallmark of puberty is the luteinizing hormone (LH) surge that triggers ovulation. Puberty initiates estrogen positive feedback onto hypothalamic circuits, which underlie the stimulation of gonadotropin releasing hormone (GnRH) neurons. In reproductively mature female rodents, both estradiol
Enhancing Effects of beta-Estradiol 3-Benzoate but Not Methoxychlor on the Promotion/Progression Stage of Chemically-induced Mammary Carcinogenesis in Ovariectomized Rats
Ueda M, et al.
Japanese Journal of Cancer Research, 93(7), 752-759 (2002)
Acupuncture does not ameliorate metabolic disturbances in the P450 aromatase inhibitor-induced rat model of polycystic ovary syndrome
Maliqueo M, et al.
Experimental Physiology, 102(1), 113-127 (2017)

Questions

  1. How should stock solutions for the product β-Estradiol 3-benzoate (E8515) be stored?

    1 answer
    1. Once the solution is prepared, it should be stored in separate packages to prevent product failure due to repeated freezing and thawing. The recommended storage period for the stock solution is as follows: when stored at -80°C, it should be used within 6 months; when stored at -20°C, it should be used within 1 month. To enhance solubility, the tube can be heated to 37°C and then agitated in an ultrasonic bath for a period of time.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service