Skip to Content
Merck
All Photos(1)

Key Documents

E1600000

Estradiol benzoate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

β-Estradiol 3-benzoate, 1,3,5(10)-Estratriene-3,17β-diol 3-benzoate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 3-benzoate, 3-Benzoyloxy-17β-estrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28O3
CAS Number:
Molecular Weight:
376.49
Beilstein:
3107526
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

estradiol

manufacturer/tradename

EDQM

mp

191-198 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)CC[C@@]3([H])[C@]1([H])CCc4cc(OC(=O)c5ccccc5)ccc24

InChI

1S/C25H28O3/c1-25-14-13-20-19-10-8-18(28-24(27)16-5-3-2-4-6-16)15-17(19)7-9-21(20)22(25)11-12-23(25)26/h2-6,8,10,15,20-23,26H,7,9,11-14H2,1H3/t20-,21-,22+,23+,25+/m1/s1

InChI key

UYIFTLBWAOGQBI-BZDYCCQFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Used in a study of general steroid metabolism and clearing, as groundwork for anabolic steroid screening in athletes.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María-Esther Cruz et al.
Alcoholism, clinical and experimental research, 38(6), 1611-1621 (2014-05-14)
Intragastric or intraperitoneal ethanol (EtOH) treatment inhibits reproductive functions in females and male rats. The area of the hypothalamus where these effects take place is unknown. As the participations of the preoptic-anterior hypothalamic area (POA-AHA) in regulating ovulation is asymmetric
Keiko Arakawa et al.
Neuroendocrinology, 100(2-3), 162-177 (2014-10-11)
Exposure to stressors such as foot shock (FS) leads to increased expression of multiple inflammatory factors, including the proinflammatory cytokine interleukin-1 (IL-1) in the brain. Studies have indicated that there are sex differences in stress reactivity, suggesting that the fluctuations
Jesús Olayo-Lortia et al.
The journal of sexual medicine, 11(10), 2428-2438 (2014-07-24)
The multiple partner choice arena (MPCA) is an experimental setup in which male rats display a significant shortening of ejaculation latency, which is the main characteristic of premature ejaculation (PE) in men. Thus, the MPCA is a potential animal model
Kara J Connelly et al.
Endocrinology, 156(1), 193-202 (2014-10-21)
Peak bone mass, one of the most important predictors for fracture risk later in life, is attained during puberty and adolescence and influenced by neonatal and pubertal sex-specific gonadal hormones and GH-IGF-I secretion patterns. This study examined the effects of
Patricia J Allen et al.
Pharmacology, biochemistry, and behavior, 130, 22-33 (2015-01-07)
Creatine is an antioxidant, neuromodulator and key regulator of energy metabolism shown to improve depressive symptoms in humans and animals, especially in females. To better understand the pharmacological effects of creatine, we examined its influence on depression-related hippocampal gene expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service