C2154
CAPSO sodium salt
Synonym(s):
3-(Cyclohexylamino)-2-hydroxy-1-propanesulfonic acid sodium salt
About This Item
Recommended Products
Assay
≥98% H2SO4 basis (anhydrous, titration)
Quality Level
form
crystalline powder
useful pH range
8.9-10.3
pKa (25 °C)
9.6
solubility
water: 0.143 g/mL, clear, colorless
application(s)
diagnostic assay manufacturing
SMILES string
[Na+].OC(CNC1CCCCC1)CS([O-])(=O)=O
InChI
1S/C9H19NO4S.Na/c11-9(7-15(12,13)14)6-10-8-4-2-1-3-5-8;/h8-11H,1-7H2,(H,12,13,14);/q;+1/p-1
InChI key
GGPORGHJKBOGDV-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service