Skip to Content
Merck
All Photos(5)

Key Documents

C2885

Sigma-Aldrich

CHES

≥99.0% (titration)

Synonym(s):

2-(Cyclohexylamino)ethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NO3S
CAS Number:
Molecular Weight:
207.29
Beilstein:
2967601
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99.0% (titration)

form

crystalline

pH

3.0-5.0 (103.6 g/L)

useful pH range

8.6-10.0

pKa (25 °C)

9.3

solubility

water: 0.6 g/25 mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

OS(=O)(=O)CCNC1CCCCC1

InChI

1S/C8H17NO3S/c10-13(11,12)7-6-9-8-4-2-1-3-5-8/h8-9H,1-7H2,(H,10,11,12)

InChI key

MKWKNSIESPFAQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CHES (2-(N-cyclohexylamino)ethanesulphonic acid) is regarded as Good′s buffers for its self-buffering and biocompatible feature. It is used in protein stabilization and does not interact with metals.

Application

CHES has been used:
  • as a buffer in mannanase activity assay of antarctic fungal isolates
  • as a buffer for DNA digestion and labeling of 2′-deoxyadenosine 3′-monophosphate
  • as a component of Wilson and Horne extraction buffer for characterization of rat plasma enzymes

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of folate in some cereals and commercial cereal-grain products consumed in Poland using trienzyme extraction and high-performance liquid chromatography methods
Gujska E and Kuncewicz A
European Food Research and Technology, 221(1-2), 208-213 (2005)
Hemicellulase activity of antarctic microfungi
Bradner JR, et al.
Journal of Applied Microbiology, 87(3), 366-370 (1999)
Good's buffers as a basis for developing self-buffering and biocompatible ionic liquids for biological research
Taha M, et al.
Green Chemistry, 16(6), 3149-3159 (2014)
Detection of Bulky Endogenous Oxidative DNA Lesions Derived from 8, 5?-Cyclo-2?-deoxyadenosine by 32P-Postlabeling Assay
Zhou GD and Moorthy B
Current Protocols in Toxicology / Editorial Board, Mahin D. Maines (editor-in-chief) ... [Et al.], 64(1), 17-17 (2015)
David G McLaren et al.
Electrophoresis, 24(17), 2887-2895 (2003-09-16)
A systematic evaluation of continuous flow-counterbalanced capillary electrophoresis for microscale preparative applications is presented. The success of the technique was found to depend on the specific nature of the analyte and background electrolyte and was most heavily influenced by factors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service