Skip to Content
Merck
All Photos(7)

Key Documents

323586

Sigma-Aldrich

Dioctyl sulfosuccinate sodium salt

≥97%

Synonym(s):

AOT, Bis(2-ethylhexyl) sulfosuccinate sodium salt, DOSS, Docusate sodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CH2O2CCH2CH(SO3Na)CO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
444.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

form

solid

mp

173-179 °C (lit.)

functional group

ester

SMILES string

[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

InChI

1S/C20H38O7S.Na/c1-5-9-11-16(7-3)14-26-19(21)13-18(28(23,24)25)20(22)27-15-17(8-4)12-10-6-2;/h16-18H,5-15H2,1-4H3,(H,23,24,25);/q;+1/p-1

InChI key

APSBXTVYXVQYAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Dioctyl sulfosuccinate sodium salt (DOSS) can be used as an anionic surfactant:
  • To prepare microemulsion with sodium salt of 3-(cyclohexylamino)-2-hydroxy-1-propanesulfonic acid (CAPSO) for the electrophoresis detection of natural and synthetic estrogens.
  • To develop reverse micelles.
  • To enhance the electrical conductivity and cell attachment in polycaprolactone fumarate and polypyrrole (PCLF–PPy) composite materials.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kagaku Kogyo, 55, 263-267 (2004)
Cutinase-AOT interactions in reverse micelles: the effect of 1-hexanol
Melo EP, et al.
Chemistry and Physics of Lipids, 124(1), 37-47 (2003)
The development of electrically conductive polycaprolactone fumarate-polypyrrole composite materials for nerve regeneration
Runge MB, et al.
Biomaterials, 31(23), 5916-5926 (2010)
Valeria Tripodi et al.
Electrophoresis, 27(22), 4431-4438 (2006-11-23)
A novel microemulsion based on sodium bis(2-ethylhexyl) sulfosuccinate (AOT) was developed for the simultaneous determination of natural and synthetic estrogens by microemulsion EKC (MEEKC). The microemulsion system consisted of 1.4% w/w AOT, 1.0% w/w octane, 7.0% w/w 1-butanol and 90.6%
Nicole Kleger et al.
Advanced materials (Deerfield Beach, Fla.), 31(37), e1903783-e1903783 (2019-07-30)
Porosity is an essential feature in a wide range of applications that combine light weight with high surface area and tunable density. Porous materials can be easily prepared with a vast variety of chemistries using the salt-leaching technique. However, this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service