Skip to Content
Merck
All Photos(1)

Key Documents

C0984

Sigma-Aldrich

Creatine Kinase MB Fraction from human heart

≥70% (SDS-PAGE), liquid

Synonym(s):

CK MB Fraction

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

Pricing and availability is not currently available.

biological source

human heart

Quality Level

Assay

≥70% (SDS-PAGE)

form

liquid

concentration

≥0.5 mg/mL

technique(s)

IR spectroscopy: suitable

suitability

suitable for tissue

UniProt accession no.

application(s)

pharmaceutical

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PHR1613PHR1504PHR3211
application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

agency

traceable to USP 1365065

agency

traceable to Ph. Eur. L0570000, traceable to USP 1365000

agency

traceable to USP 1368008

agency

traceable to USP 1448989

API family

levothyroxine

API family

levothyroxine

API family

liothyronine

API family

mycophenolate

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

packaging

pkg of 1 g

packaging

pkg of 500 mg

packaging

pkg of 400 mg

General description

Research area: Cell Signaling
Creatine Kinase (CK) comprises M and B subunit and is a dimeric molecule. It is one among the isoenzymes of CK. The enzyme is majorly present in the myocardium.[1]

Application

Creatine Kinase MB Fraction from human heart has been used:
  • as a diagnostic marker for acute myocardial infarction.[2]
  • to identify overlapping absorbance peaks between blood and the cardiac biomarkers.[3]
  • to study the phosphorylation pathway of dioxolane guanosine (DXG) in vivo, a potential treatment of HIV-1 infection.[4]

Biochem/physiol Actions

Creatine Kinase (CK) catalyzes the formation of creatinine from creatine phosphate.[1] The enzyme is associated with coronary performance. Elevated serum levels of CK-MB are prime indicators of myocardial cellular injury.[1] It is regarded as a “gold standard” for myocardial infarction diagnosis.[1] The levels of cardiac troponin I (cTnI) or cardiac troponin T also correlate to higher CK-MB) levels.[5] CK-MB levels may also be elevated in chronic renal failure.[6]

Components

Solution in 5 mM sodium succinate, 10 mM sodium chloride, 1 mM EDTA, 5 mM β-mercaptoethanol, 50% glycerol, pH 7.0.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Need A Sample COA?

    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Development and preliminary validation of infrared spectroscopic device for transdermal assessment of elevated cardiac troponin
    Titus J, et al.
    Communications medicine, 2 (2022)
    J E Adams et al.
    Clinical chemistry, 40(7 Pt 1), 1291-1295 (1994-07-01)
    Although measurement of cardiac troponin I (cTnI) is, in some situations, more specific for detection of cardiac injury than is measurement of the MB isoenzyme of creatine kinase (MBCK), its sensitivity and specificity relative to MBCK for detection of myocardial
    Terry M Welsh et al.
    Annals of clinical and laboratory science, 32(1), 44-49 (2002-02-19)
    The diagnosis of myocardial infarction (MI) is established in patients with chest pain and equivocal electrocardiogram changes by demonstrating a rise in blood levels of creatine kinase MB (CK-MB) and/or an increase in cardiac troponin I (cTnI) or cardiac troponin
    Anabolism of amdoxovir: phosphorylation of dioxolane guanosine and its 5?-phosphates by mammalian phosphotransferases
    Feng JY, et al.
    Biochemical Pharmacology, 68(9), 1879-1888 (2004)
    Creatine Kinase
    Clinical Methods: The History, Physical, and Laboratory Examinations (1990)

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service