PHR2880
Levothyroxine Sodium Hydrate
pharmaceutical secondary standard, certified reference material
Synonym(s):
Levothyroxine sodium hydrate, 3,3′,5,5′-Tetraiodo-L-thyronine monosodium salt hydrate, L-Thyroxin monosodium salt hydrate, Sodium O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-L-tyrosine hydrate
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1365065
API family
levothyroxine
CofA
current certificate can be downloaded
packaging
pkg of 100 mg
application(s)
pharmaceutical
storage temp.
2-30°C
SMILES string
O.[Na+].N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C([O-])=O
InChI
1S/C15H11I4NO4.Na.H2O/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23;;/h1-2,4-5,12,21H,3,20H2,(H,22,23);;1H2/q;+1;/p-1/t12-;;/m0../s1
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | PHR1613 | PHR1504 | PHR3211 |
---|---|---|---|
pharmaceutical | pharmaceutical (small molecule) | pharmaceutical (small molecule) | pharmaceutical |
traceable to USP 1365065 | traceable to Ph. Eur. L0570000, traceable to USP 1365000 | traceable to USP 1368008 | traceable to USP 1448989 |
levothyroxine | levothyroxine | liothyronine | mycophenolate |
current certificate can be downloaded | current certificate can be downloaded | current certificate can be downloaded | current certificate can be downloaded |
pkg of 100 mg | pkg of 1 g | pkg of 500 mg | pkg of 400 mg |
General description
Application
Analysis Note
Other Notes
Recommended products
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 1
Target Organs
Thyroid,Cardio-vascular system,Kidney
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service