Skip to Content
Merck
All Photos(3)

Key Documents

MAK055

Sigma-Aldrich

AST Activity Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Aspartate Aminotransferase Activity Assay Kit, Glutamate-Oxaloacetate Transaminase Activity Assay Kit, SGOT Activity Assay Kit, Serum Glutamate-Oxaloacetate Transaminase Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

gastrointestinal diseases; cardiovascular diseases

storage temp.

−20°C

Gene Information

human ... GOT1(2805)
mouse ... GOT1(14718)
rat ... GOT1(24401)

General description

The new AST Assay Kit, MAK467, is now available! Aspartate Aminotransferase (AST), formerly called serum glutamate-oxaloacetate transaminase (SGOT), is a pyridoxal phosphate (PLP)-dependent enzyme that catalyzes the conversion of aspartate and α-ketoglutarate to oxaloacetate and glutamate. Similar to Alanine Aminotransferase (ALT), AST levels in blood are commonly used as a marker for liver function. However, AST has a broader tissue distribution than ALT and perturbations in AST levels can occur in response to diseases or injuries in multiple tissues including skeletal and heart.

Application

AST Activity Assay Kit has been used to determine the activity of aspartate aminotransferase in samples.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the measurement of aspartate aminotransferase (AST) activity in biological samples including tissue, cells and serum

Principle

The Aspartate Aminotransferase Activity Assay kit provides a simple and direct procedure for measuring AST activity in a variety of samples. In this kit, the transfer of an amino group from aspartate to α-ketoglutarate results in the generation of glutamate, resulting in the production of a colorimetric (450 nm) product proportional to the AST enzymatic activity present. One unit of AST is the amount of enzyme that will generate 1.0 μmole of glutamate per minute at pH 8.0 at 37 °C.

replaced by

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leola N Chow et al.
PloS one, 11(3), e0151765-e0151765 (2016-03-22)
Modulation of chemokine CXCL12 and its receptor CXCR4 has been implicated in attenuation of bleomycin (BLM)-induced pulmonary fibrosis and carbon tetrachloride (CCl4)-induced hepatic injury. In pulmonary fibrosis, published reports suggest that collagen production in the injured lung is derived from
Tiffany A Katz et al.
Environmental health perspectives, 128(1), 17010-17010 (2020-01-16)
Tributyltin (TBT) is a persistent and bioaccumulative environmental toxicant. Developmental exposure to TBT has been shown to cause fatty liver disease (steatosis), as well as increased adiposity in many species, leading to its characterization as an obesogen. We aimed to
Xinyi Xu et al.
Microbiology (Reading, England), 161(6), 1219-1230 (2015-03-27)
Aspartate and glutamate are two key amino acids used in biosynthesis of many amino acids that play vital role in cellular metabolism. Aspartate aminotransferases (AspATs) are required for channelling nitrogen (N(2)) between Glu and Asp in all life forms. Biochemical
Resolvin D1 and E1 alleviate the progress of hepatitis toward liver cancer in long-term concanavalin A-induced mice through inhibition of NF-KB activity.
Kuang H, et al.
Oncology Reports, 35, 307-317 (2016)
Zhiqiang Ye et al.
Journal of cellular biochemistry, 119(9), 7397-7405 (2018-05-19)
Liver ischemia/reperfusion (I/R) injury has high mortality due to the intense inflammatory process occurs in the liver. However, the pathological mechanism underlying I/R injury is still not clear. Recent works showed that circular RNAs play critical roles in many human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service