Skip to Content
Merck
All Photos(1)

Key Documents

AV36019

Sigma-Aldrich

Anti-EBF2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-COE2, Anti-EBF-2, Anti-Early B-cell factor 2, Anti-FLJ11500, Anti-O/E-3, Anti-OE-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

61 kDa

species reactivity

human, rat, rabbit, guinea pig, horse, mouse, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EBF2(64641)

Related Categories

Immunogen

Synthetic peptide directed towards the C terminal region of human EBF2

Biochem/physiol Actions

EBF2 belongs to the EBF/COE family of transcription factors that contain well-conserved DNA-binding domain. EBF factors play important regulatory roles in various developmental processes and in differentiation of osteoblasts. EBF2 is critical in the generation and migration of Cajal-Retzius neurons during early cortical neurogenesis in cerebral cortex.

Sequence

Synthetic peptide located within the following region: VPRNPSQLPALSSSPAHSGMMGINSYGSQLGVSISESTQGNNQGYIRNTS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Chiara et al.
Developmental biology, 365(1), 277-289 (2012-03-17)
Cajal-Retzius (CR) cells play a crucial role in the formation of the cerebral cortex, yet the molecules that control their development are largely unknown. Here, we show that Ebf transcription factors are expressed in forebrain signalling centres-the septum, cortical hem
Shu-Mien Chuang et al.
Developmental neuroscience, 33(6), 479-493 (2011-11-02)
Mammalian cortical neurogenesis occurs on a precise time schedule during development. The earliest born neurons form the preplate and later separate into layer 1, which includes Cajal-Retzius (C-R) neurons, and the subplate. The preplate and its derivatives play a critical
Ana Patiño-García et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 15(16), 5082-5091 (2009-08-13)
Osteosarcoma is the most prevalent bone tumor in children and adolescents. At present, the mechanisms of initiation, maintenance, and metastasis are poorly understood. The purpose of this study was to identify relevant molecular targets in the pathogenesis of osteosarcoma. Tumor
S S Wang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 17(11), 4149-4158 (1997-06-01)
The Olf-1/EBF helix-loop-helix (HLH) transcription factor has been implicated in olfactory gene regulation and in B-cell development. Using homology screening methods, we identified two additional Olf-1/EBF-like cDNAs from a mouse embryonic cDNA library. The Olf-1/EBF-like (O/E) proteins O/E-1, O/E-2, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service