Skip to Content
Merck
All Photos(1)

Key Documents

70584-M

Millipore

BugBuster® Protein Extraction Reagent

Synonym(s):

Protein Extraction Reagent, Protein Extraction Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106516
NACRES:
NA.32

form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

10-30°C

General description

BugBuster® Protein Extraction Reagent is formulated for gentle disruption of Escherichia coli cell wall to liberate active proteins. It provides a simple, rapid, low-cost alternative to mechanical methods such as French Press or sonication for releasing expressed target protein in preparation for purification or other applications. The proprietary formulation utilizes a tris-buffer-based mixture of non-ionic and zwitterionic detergents that is capable of cell wall perforation without denaturing the protein. BugBuster is also useful for the preparation of high-purity inclusion bodies for subsequent purification in instances where expressed proteins are insoluble.

Application

BugBuster® Protein Extraction Reagent has been used:
  • to lyse Escherichia coli BLR (DE3) cells for β-glucosidase (BglB) production and purification
  • as a cell lysis buffer to treat the synechocystis PCC6803 transconjugants and measure the yellow fluorescent protein (YFP) fluorescence
  • in lysis buffer for purification of glutathione S-transferase- heat shock protein family A (Hsp70) member 8 (GST- HSPA8)

Warning

Toxicity: Standard Handling (A)

Legal Information

BUGBUSTER is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dylan Alexander Carlin et al.
PloS one, 12(5), e0176255-e0176255 (2017-05-23)
Accurate modeling of enzyme activity and stability is an important goal of the protein engineering community. However, studies seeking to evaluate current progress are limited by small data sets of quantitative kinetic constants and thermal stability measurements. Here, we report
Patricia Armshaw et al.
Biotechnology for biofuels, 8, 201-201 (2015-12-03)
The use of photosynthetic autotrophs and in particular the model organism Synechocystis PCC6803 is receiving much attention for the production of sustainable biofuels and other economically useful products through metabolic engineering. Optimisation of metabolic-engineered organisms for high-level sustained production of
Tiejian Nie et al.
Autophagy, 17(10), 2923-2938 (2021-05-11)
As a highly dynamic organelle, mitochondria undergo constant fission and fusion to change their morphology and function, coping with various stress conditions. Loss of the balance between fission and fusion leads to impaired mitochondria function, which plays a critical role

Articles

Citation Spotlight: BugBuster® Protein Extraction Reagent for Efficient Protein Extraction from Bacterial Pathogens

This article shows the use of BugBuster® and Benzonase® as protein purification tools to extract recombinant proteins from E. coli and to reduce the viscosity of the extract.

This article shows the use of BugBuster® and Benzonase® as protein purification tools to extract recombinant proteins from E. coli and to reduce the viscosity of the extract.

This article shows the use of BugBuster® and Benzonase® as protein purification tools to extract recombinant proteins from E. coli and to reduce the viscosity of the extract.

See All

Related Content

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service