A6251
N-Acetyl-DL-tryptophan
≥98% (TLC)
Synonym(s):
N-acetyl-tryptophan
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H14N2O3
CAS Number:
Molecular Weight:
246.26
Beilstein:
89478
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
N-Acetyl-DL-tryptophan,
Assay
≥98% (TLC)
Quality Level
form
powder
mp
204-206 °C (dec.) (lit.)
storage temp.
2-8°C
SMILES string
CC(=O)NC(Cc1c[nH]c2ccccc12)C(O)=O
InChI
1S/C13H14N2O3/c1-8(16)15-12(13(17)18)6-9-7-14-11-5-3-2-4-10(9)11/h2-5,7,12,14H,6H2,1H3,(H,15,16)(H,17,18)
InChI key
DZTHIGRZJZPRDV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Mitigation of Oxidation in Therapeutic Antibody Formulations: a Biochemical Efficacy and Safety Evaluation of N-Acetyl-Tryptophan and L-Methionine.: Investigates N-Acetyl-DL-tryptophan′s role in reducing oxidative damage in antibody formulations, enhancing therapeutic efficacy (Dion et al., 2018).
- Effect of PEGylation on performance of protein microbubbles and its comparison with lipid microbubbles.: Examines how PEGylation, in combination with N-Acetyl-DL-tryptophan, improves the stability and performance of protein microbubbles in biomedical applications (Upadhyay et al., 2017).
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Takahiro Takayama et al.
Analytica chimica acta, 898, 73-84 (2015-11-04)
Chiral metabolites are found in a wide variety of living organisms and some of them are understood to be physiologically active compounds and biomarkers. However, the overall analysis of chiral metabolomics is quite difficult due to the high number of
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service