Skip to Content
Merck
All Photos(3)

Key Documents

M3148

Sigma-Aldrich

2-Mercaptoethanol

for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)

Synonym(s):

β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2OH
CAS Number:
Molecular Weight:
78.13
Beilstein:
773648
EC Number:
MDL number:
UNSPSC Code:
12161504
PubChem Substance ID:
NACRES:
NA.31

grade

for molecular biology

Quality Level

vapor density

2.69 (vs air)

vapor pressure

1 mmHg ( 20 °C)

product line

BioReagent

Assay

99% (GC/titration)

form

liquid

expl. lim.

18 %

reaction suitability

reagent type: reductant

concentration

14.3 M (pure liquid)

technique(s)

cell culture | mammalian: suitable
electrophoresis: suitable

refractive index

n20/D 1.500 (lit.)

pH

4.5-6 (20 °C, 500 g/L)

bp

157 °C (lit.)

solubility

H2O: soluble 1 mL/mL

density

1.114 g/mL at 25 °C (lit.)

foreign activity

DNase, RNase, protease, none detected

storage temp.

2-8°C

SMILES string

OCCS

InChI

1S/C2H6OS/c3-1-2-4/h3-4H,1-2H2

InChI key

DGVVWUTYPXICAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reducing agent
β-mercaptoethanol or 2-mercaptoethanol has been used:
  • as a supplement in Roswell Park Memorial Institute (RPMI)-1640 medium to culture DT40 cells (chicken B cell line)
  • to lyse NuLi-1 (Normal Lung, University of Iowa-1) cells
  • to trypsinize mouse embryonic stem cells
BME is suitable for reducing protein disulfide bonds prior to polyacrylamide gel electrophoresis and is usually included in a sample buffer for SDS-PAGE at a concentration of 5%. Cleaving intermolecular (between subunits) disulfide bonds allows the subunits of a protein to separate independently on SDS-PAGE. Cleaving intramolecular (within subunit) disulfide bonds allows the subunits to become completely denatured so that each peptide migrates according to its chain length with no influence due to secondary structure.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 2 Oral

Target Organs

Liver,Heart

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced Innate Immune Response to a Staphylococcus aureus Small Colony Variant Compared to Its Wild-Type Parent Strain
Judy J
Frontiers in Cellular and Infection Microbiology, 6, 187-187 (2016)
A subset of platinum-containing chemotherapeutic agents kills cells by inducing ribosome biogenesis stress
Peter M Bruno
Nature Medicine, 23 (2017)
Yasunari Seita et al.
Biology of reproduction, 100(6), 1440-1452 (2019-03-15)
Nonhuman primates (NHPs) are considered to be the most valuable models for human transgenic (Tg) research into disease because human pathology is more closely recapitulated in NHPs than rodents. Previous studies have reported the generation of Tg NHPs that ubiquitously
Kotaro Fujii et al.
Nature communications, 8, 14443-14443 (2017-02-15)
The degree and dynamics of translational control during mammalian development remain poorly understood. Here we monitored translation of the mammalian genome as cells become specified and organize into tissues in vivo. This identified unexpected and pervasive translational regulation of most
Sheila R Costford et al.
American journal of physiology. Endocrinology and metabolism, 306(12), E1341-E1353 (2014-04-25)
Chronic low-grade inflammation is an important contributor to the development of insulin resistance, a hallmark of type 2 diabetes mellitus (T2DM). Obesity and high-fat feeding lead to infiltration of immune cells into metabolic tissues, promoting inflammation and insulin resistance. We

Protocols

XTT assay protocol for measuring cell viability, proliferation, activation and cytotoxicity. Instructions for XTT reagent preparation and examples of applications.

MTT assay protocol for measuring cell viability, proliferation and cytotoxicity. Instructions for MTT reagent preparation and examples of applications.

MTT assay protocol for measuring cell viability, proliferation and cytotoxicity. Instructions for MTT reagent preparation and examples of applications.

MTT assay protocol for measuring cell viability, proliferation and cytotoxicity. Instructions for MTT reagent preparation and examples of applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service