Skip to Content
Merck
All Photos(1)

Key Documents

441554

Sigma-Aldrich

Nα-Acetyl-L-asparagine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
174.15
Beilstein:
1726196
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

mp

168-170 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(=O)N[C@@H](CC(N)=O)C(O)=O

InChI

1S/C6H10N2O4/c1-3(9)8-4(6(11)12)2-5(7)10/h4H,2H2,1H3,(H2,7,10)(H,8,9)(H,11,12)/t4-/m0/s1

InChI key

HXFOXFJUNFFYMO-BYPYZUCNSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Regulatory role of the excitatory amino acid system in the neuromediator response of the hypothalamus to stress].
E I Chazov et al.
Doklady Akademii nauk, 351(4), 567-570 (1996-12-01)
T M Agaev et al.
Voprosy meditsinskoi khimii, 37(4), 37-40 (1991-07-01)
Under conditions of 45-days visual deprivation content of N-acetyl-L-aspartic acid was considerably increased in visual cortex (field 17) tissues and decreased in the superior colliculus and exterior geniculate body. At the same time, content of N-acetyl-L-aspartic acid was not altered
31P-MR spectroscopy in monozygotic twins discordant for schizophrenia or schizoaffective disorder.
Igor Nenadic et al.
Schizophrenia research, 134(2-3), 296-297 (2011-11-05)
Suja Saraswathy et al.
Journal of neuro-oncology, 91(1), 69-81 (2008-09-24)
Purpose Glioblastoma Multiforme (GBM) is the most common and lethal primary brain tumor in adults. The goal of this study was to test the predictive value of MR parameters in relation to the survival of patients with newly diagnosed GBM
David N Guilfoyle et al.
NMR in biomedicine, 16(8), 468-474 (2003-12-30)
An inverse relationship between applied magnetic field strength and the apparent diffusion coefficient (ADC) of several important brain metabolites including N-acetyl-l-aspartate (NAA), choline and creatine, measured in vivo using proton magnetic resonance spectroscopy (MRS), has been reported. In this investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service