Skip to Content
Merck
All Photos(1)

Key Documents

A3187

Sigma-Aldrich

Anti-Human IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Goat Anti-Human IgG (γ-chain specific)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

direct ELISA: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders . Anti-Human IgG (γ-chain specific)-Alkaline Phosphatase antibody is specific for human IgG when tested against human IgA, IgG, IgM, and Bence Jones κ, and λ myeloma proteins.

Immunogen

Purified human IgG

Application

Anti-Human IgG (γ-chain specific)-Alkaline Phosphatase antibody is suitable for use in ELISA (at 1:2500 and 1:10,000 dilutions). The antibody can also be used for western blot (1:30,000) and immunohistochemistry (at 1:50 dilutions using formalin-fixed, paraffin-embedded sections).
Proteins harvested from E. coli cultures were run on SDS-page gel for western blot analysis using alkaline phosphatase conjugated goat anti-human IgG (chain specific) at a concentration of 1:10000 for the secondary. IgG was detected in human serum by Elisa using alkaline phosphatase conjugated goat anti-human IgG (gamma-chain specific) diluted 1:5000 in 0.25% BSA/PBS. Assay was developed using p-nitrophenyl phosphate substrate (1mg/ml Sigma).

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 10 mM glycine, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Akyol et al.
Seizure, 16(3), 233-237 (2007-01-24)
Increased seropositivity for Toxoplasma gondii and Toxocara canis have been observed in epileptic patients. Our aim is to determine whether there is any relationship between these agents and epilepsy in our cryptogenic epilepsy group. We studied specific IgG antibodies against
Rodrigo Lorenzi et al.
Annals of clinical biochemistry, 51(Pt 2), 248-257 (2013-08-29)
The soluble form of the receptor for advanced glycation end-products (sRAGE) has been studied in various diseases. It is not clear why sRAGE levels vary between studies, with controversial results. What also remains to be determined is whether receptor for
D A Heim et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 1(6), 533-544 (2000-08-10)
Host immune responses against foreign transgenes may be a major obstacle to successful gene therapy. To clarify the impact of an immune response to foreign transgene products on the survival of genetically modified cells, we studied the in vivo persistence
Meng-Yun Chou et al.
The Journal of clinical investigation, 119(5), 1335-1349 (2009-04-14)
Atherosclerosis is a chronic inflammatory disease characterized by the accumulation of oxidized lipoproteins and apoptotic cells. Adaptive immune responses to various oxidation-specific epitopes play an important role in atherogenesis. However, accumulating evidence suggests that these epitopes are also recognized by
Assaf Raz et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(18), 4781-4786 (2017-04-22)
The cell wall of Gram-positive bacteria contains abundant surface-exposed carbohydrate molecules that are highly conserved within and often across species. The potential therapeutic usefulness of high-affinity antibodies to cell wall carbohydrates is unquestioned, however obtaining such antibodies is challenging due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service