Skip to Content
Merck
All Photos(1)

Key Documents

A3150

Sigma-Aldrich

Anti-Human IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Goat Anti-Human IgG (γ-chain specific)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

direct ELISA: 1:7,000-1:21,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Immunoglobulins (Igs) belongs to the immunoglobulin super-family. Each immunoglobin has two heavy (H) and two light (L) chains held together by disulfide linkages. Each light chain comprises one variable N-terminal region and a constant C-terminal region. Heavy chain has one variable N-terminal region and three or four constant (CH1-CH4) C-terminal region. IgG is a glycoprotein antibody that contains two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4.

Immunogen

purified human IgG

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Enzyme-linked immunosorbent assay (1 paper)
Goat anti-human IgG (gamma-chain specific)-alkaline phosphatase antibody can be used for ELISA applications using serum samples from allergic and borderline hypertensive patients.
Proteins harvested from E. coli cultures were run on SDS-page gel for western blot analysis using alkaline phosphatase conjugated goat anti-human IgG (chain specific) at a concentration of 1:10000 for the secondary. IgG was detected in human serum by Elisa using alkaline phosphatase conjugated goat anti-human IgG (gamma-chain specific) diluted 1:5000 in 0.25% BSA/PBS. Assay was developed using p-nitrophenyl phosphate substrate (1mg/ml Sigma).

Biochem/physiol Actions

Human IgGs regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders. Goat anti-human IgG (gamma-chain specific)-alkaline phosphatase antibody is specific for human IgG when tested against human IgA, IgG, IgM, Bence Jones Kappa, and Lambda myeloma proteins.
IgG1 is most abundant and its deficiency results in hypogammaglobulinemia. IgG2 deficiency increases susceptibility to bacterial infections. IgG3 mediates effector functions. IgG4 is associated with asymptomatic infection. IgG antibody have enormous therapeutic potential and the Fc region aids in the development of therapeutic antibody.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2, 15 mM sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Hühn et al.
Nucleic acids research, 25(2), 410-416 (1997-01-15)
During the analysis of the La (SS-B) autoantigen for catalytic activities an ATP-dependent double-stranded RNA unwinding activity was detected. Both native and recombinant La proteins from different species displayed this activity, which could be inhibited by monospecific anti-La antibodies. La
X Baur et al.
Allergy, 57(10), 943-945 (2002-09-25)
Although fungal phytase is frequently used as an additive to animal feed few investigations of its allergenic property have been conducted. Fifty-three subjects occupationally exposed to powdered phytase from Aspergillus niger were studied. Exposure data and symptoms were registered by
K P Ng et al.
Arthritis and rheumatism, 55(6), 900-904 (2006-12-02)
To identify the prevalence of serologically active clinically quiescent (SACQ) patients in a cohort of 290 patients with systemic lupus erythematosus (SLE). We investigated if the presence of anti-double-stranded DNA (anti-dsDNA) or antinucleosome (anti-NCS) antibodies during the SACQ period was
R Wu et al.
Hypertension (Dallas, Tex. : 1979), 33(1), 53-59 (1999-02-04)
-Elevated antibody levels to oxidized low-density lipoprotein (aOxLDL) have been shown to correlate with the degree of atherosclerosis in some studies. On the other hand, immunization of experimental animals with OxLDL, leading to enhanced aOxLDL levels, inhibits the development of
H Tröster et al.
Journal of autoimmunity, 8(6), 825-842 (1995-12-01)
Immunization of Balb/c mice with a homogeneously purified recombinant human La/SS-B protein resulted in activation of an autoreactive B cell secreting a novel monoclonal anti-La antibody termed La4B6. La4B6 reacted with La protein from a variety of sources including human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service