Skip to Content
Merck
All Photos(1)

Key Documents

A6029

Sigma-Aldrich

Anti-Human IgG (γ-chain specific)−Peroxidase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Human IgG (γ-chain specific)−HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

direct ELISA: 1:10,000

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders. Goat Anti-Human IgG (γ-chain specific)-Peroxidase antibody is specific for human IgG when tested against human IgA, IgG, IgM, Bence Jones Kappa and Lambda myeloma proteins.

Immunogen

purified human IgG

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Enzyme-linked immunosorbent assay (1 paper)
Goat Anti-Human IgG (γ-chain specific)-Peroxidase antibody has been used for several ELISA-based applications.
Human serum samples were analyzed for the presence of anti-hepatitis B viral surface antigens by ELISA using HRP-conjugated goat anti-human IgG gamma chain-specific as the secondary antibody.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4 containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alejandro Luquetti Ostermayer et al.
Revista da Sociedade Brasileira de Medicina Tropical, 44 Suppl 2, 108-121 (2011-05-21)
A survey for seroprevalence of Chagas disease was held in a representative sample of Brazilian individuals up to 5 years of age in all the rural areas of Brazil, with the single exception of Rio de Janeiro State. Blood on
J Kapusta et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 13(13), 1796-1799 (1999-10-03)
The infectious hepatitis B virus represents 42 nm spherical double-shelled particles. However, analysis of blood from hepatitis B virus carriers revealed the presence of smaller 22 nm particles consisting of a viral envelope surface protein. These particles are highly immunogenic
Sandeep Gupta et al.
PLoS pathogens, 9(11), e1003776-e1003776 (2013-11-28)
The mechanisms by which human immunodeficiency virus type 1 (HIV-1) crosses mucosal surfaces to establish infection are unknown. Acidic genital secretions of HIV-1-infected women contain HIV-1 likely coated by antibody. We found that the combination of acidic pH and Env-specific
J S Crowe et al.
Clinical and experimental immunology, 87(1), 105-110 (1992-01-01)
Expression of CAMPATH-1H, a humanized MoAb directed against an abundant surface antigen on human lymphocytes, has been studied using transfected myeloma cells. As the site of chromosome integration of DNA transfected into a cell is random we investigated the feasibility
Khojasteh V Javid et al.
Iranian journal of microbiology, 3(4), 170-176 (2012-04-25)
The aim of study was to develop a rapid assay, dye labelled monoclonal antibody assay (DLMAA), using non-radioactive organic synthetic dyes for identification of Toxic Shock Syndrome Toxin-1 (TSST-1) producing strains of Staphylococcus aureus. The assay protocol required only two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service