Skip to Content
Merck
All Photos(1)

Key Documents

A0773

Sigma-Aldrich

Angiotensin Converting Enzyme Inhibitor

≥95% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H76N14O12
CAS Number:
Molecular Weight:
1101.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥95% (TLC)

form

powder

mol wt

~_1.1 kDa

solubility

water: soluble 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]2CCCN2C(=O)[C@H](Cc3c[nH]c4ccccc34)NC(=O)[C@@H]5CCC(=O)N5)C(=O)N6CCC[C@H]6C(=O)N7CCC[C@H]7C(O)=O

InChI

1S/C53H76N14O12/c1-3-29(2)43(51(77)66-25-9-16-39(66)50(76)67-26-10-17-40(67)52(78)79)63-45(71)34(18-20-41(54)68)60-46(72)37-14-7-23-64(37)48(74)35(13-6-22-57-53(55)56)61-47(73)38-15-8-24-65(38)49(75)36(62-44(70)33-19-21-42(69)59-33)27-30-28-58-32-12-5-4-11-31(30)32/h4-5,11-12,28-29,33-40,43,58H,3,6-10,13-27H2,1-2H3,(H2,54,68)(H,59,69)(H,60,72)(H,61,73)(H,62,70)(H,63,71)(H,78,79)(H4,55,56,57)/t29-,33-,34-,35-,36-,37-,38-,39-,40-,43-/m0/s1

InChI key

UUUHXMGGBIUAPW-CSCXCSGISA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Bello et al.
Bioorganic & medicinal chemistry, 18(9), 3320-3334 (2010-03-30)
Novel alpha-mannosidase inhibitors of the type (2R,3R,4S)-2-({[(1R)-2-hydroxy-1-arylethyl]amino}methyl)pyrrolidine-3,4-diol have been prepared and assayed for their anticancer activities. Compound 30 with the aryl group=4-trifluoromethylbiphenyl inhibits the proliferation of primary cells and cell lines of different origins, irrespective of Bcl-2 expression levels, inducing
Of mice and men: therapeutic mTOR inhibition in polycystic kidney disease.
York Pei
Journal of the American Society of Nephrology : JASN, 21(3), 390-391 (2010-02-06)
Oscar Gallardo et al.
The Journal of biological chemistry, 285(4), 2721-2733 (2009-11-27)
Paenibacillus barcinonensis is a soil bacterium bearing a complex set of enzymes for xylan degradation, including several secreted enzymes and Xyn10B, one of the few intracellular xylanases reported to date. The crystal structure of Xyn10B has been determined by x-ray
Linqing Ma et al.
Journal of Alzheimer's disease : JAD, 27(1), 225-237 (2011-07-30)
Apomorphine hydrochloride (APO) is known to be a dopamine receptor agonist, and has recently been found to be a novel drug for Alzheimer's disease (AD). We found that APO treatment ameliorated oxidative stress in an AD mouse model and specifically
R Matsas et al.
The Biochemical journal, 223(2), 433-440 (1984-10-15)
Endopeptidase-24.11 (EC 3.4.24.11), purified to homogeneity from pig kidney, was shown to hydrolyse a wide range of neuropeptides, including enkephalins, tachykinins, bradykinin, neurotensin, luliberin and cholecystokinin. The sites of hydrolysis of peptides were identified, indicating that the primary specificity is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service